Skip to Content
Merck
All Photos(1)

Key Documents

39090

Supelco

5-(4-Dimethylaminobenzylidene)rhodanine

for TLC derivatization, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2OS2
CAS Number:
Molecular Weight:
264.37
Beilstein:
189065
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (S)
≥98.0%

technique(s)

thin layer chromatography (TLC): suitable

mp

285-288 °C (lit.)

solubility

dioxane: 0.1 g/10 mL (hot)

SMILES string

CN(C)c1ccc(cc1)\C=C2\SC(=S)NC2=O

InChI

1S/C12H12N2OS2/c1-14(2)9-5-3-8(4-6-9)7-10-11(15)13-12(16)17-10/h3-7H,1-2H3,(H,13,15,16)/b10-7+

InChI key

JJRVRELEASDUMY-JXMROGBWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

for the detection of Ag, Au, Cu, Hg, Pd, Pt, alkaloids, antipyrin, indican, sulfonamide, urobilinogen

Other Notes

Reagent for the determination of silver, palladium and mercury ; Determination of gold; Used in the photometric determination of autoradiograms

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

5 (4-dimethylaminobenzylidene) rhodanine as a reagent for the spectrophotometric determination of copper and manganese.
Ojeka, E. O., and J. F. Iyun
Global Journal of Pure and Applied Sciences, 10.4, 577-583 (2004)
G. Jangg
Fresenius Journal of Analytical Chemistry, 183, 255-255 (1961)
G.H. Ayres et al.
Analytica Chimica Acta, 24, 241-241 (1961)
L.E. Lichtenstein
Analytical Chemistry, 47, 465-465 (1975)
Reinhard Dallinger et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 289(4), R1185-R1195 (2005-05-21)
Like all other animal species, terrestrial pulmonate snails require Cu as an essential trace element. On the other hand, elevated amounts of Cu can exert toxic effects on snails. The homeostatic regulation of Cu must therefore be a pivotal goal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service