Skip to Content
Merck
All Photos(1)

Documents

36542

Supelco

DEET

PESTANAL®, analytical standard

Synonym(s):

N,N-Diethyl-m-toluamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17NO
CAS Number:
Molecular Weight:
191.27
Beilstein:
2046711
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCN(CC)C(=O)c1cccc(C)c1

InChI

1S/C12H17NO/c1-4-13(5-2)12(14)11-8-6-7-10(3)9-11/h6-9H,4-5H2,1-3H3

InChI key

MMOXZBCLCQITDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

DEET can find wide applications as an active ingredient in insect repellents, which can be used for protection against potentially disease-carrying insects and ticks such as deer ticks leading to lyme disease, mosquitoes leading to malaria, dengue fever, zika virus and west nile virus. It can also be used for protection against the condition of encephalitis inflammation.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Reaction mechanism and kinetics of DEET visible light assisted photocatalytic ozonation with WO 3 catalyst
Mena E, et al.
Applied Catalysis. B, Environmental, 202, 460-472 (2017)
Novel exposure biomarkers of N, N-diethyl-m-toluamide (DEET): Data from the 2007?2010 National Health and Nutrition Examination Survey
Calafat.MA, et al.
Environment International, 92, 398-404 (2016)
Ali Afify et al.
Malaria journal, 19(1), 127-127 (2020-04-02)
The species-specific mode of action for DEET and many other mosquito repellents is often unclear. Confusion may arise for many reasons. First, the response of a single mosquito species is often used to represent all mosquito species. Second, behavioural studies
T G Osimitz et al.
Journal of toxicology. Clinical toxicology, 35(5), 435-441 (1997-01-01)
N,N-Diethyl-m-toluamide, an effective tick repellent, when applied to the skin, is the major component of essentially all of the products marketed for this purpose. It is used by about 30% of the US population, and by 23-29% of children in
Daniel L Sudakin et al.
Journal of toxicology. Clinical toxicology, 41(6), 831-839 (2003-12-18)
The emergence of West Nile Virus (WNV) in North America has resulted in increased public awareness and utilization of insect repellents containing N,N-diethyl-m-toluamide (DEET) in the prevention of mosquito-borne disease. Regulatory reassessments in North American countries have recently been completed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service