Skip to Content
Merck
All Photos(1)

Documents

35758

Supelco

Alternariol

analytical standard

Synonym(s):

3,7,9-Trihydroxy-1-methyl-6H-dibenzo[b,d]pyran-6-one, 3,7,9-Trihydroxy-1-methyl-benzo[c]chromen-6-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10O5
CAS Number:
Molecular Weight:
258.23
Beilstein:
244839
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

Cc1cc(O)cc2OC(=O)c3c(O)cc(O)cc3-c12

InChI

1S/C14H10O5/c1-6-2-7(15)5-11-12(6)9-3-8(16)4-10(17)13(9)14(18)19-11/h2-5,15-17H,1H3

InChI key

CEBXXEKPIIDJHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Alternariol (AOH) is a mycotoxin produced by fungus Alternaria and a major plant pathogen found in a wide variety of fruits and cereals products. AOH is structurally characterized as a dibenzopyranone.

Application

Alternariol may be used as an analytical reference standard for the determination of the analyte in fruits, vegetables and cereals by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Reconstitution

Dried down, ~100μg/mL after reconstitution with 1 mL of water

Analysis Note

purity : ≥96.0% (HPLC)

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Precise determination of the Alternaria mycotoxins alternariol and alternariol monomethyl ether in cereal, fruit and vegetable products using stable isotope dilution assays.
Asam S, et al.
Mycotoxin Research, 27(1), 23-28 (2011)
A Solhaug et al.
Toxicology letters, 219(1), 8-17 (2013-03-05)
The mycotoxin alternariol (AOH), a frequent contaminant in fruit and cereal products, is known to induce DNA damage with subsequent cell cycle arrest. Here we elucidated the effects of AOH on stages of cell cycle progression using the RAW 264.7
Stefanie C Fleck et al.
Toxicology letters, 214(1), 27-32 (2012-08-21)
Altertoxin II (ATX II) is one of the several mycotoxins produced by Alternaria fungi. It has a perylene quinone structure and is highly mutagenic in Ames Salmonella typhimurium, but its mutagenicity in mammalian cells has not been studied before. Here
Eva Graf et al.
International journal of food microbiology, 157(3), 353-359 (2012-06-26)
Strains of the genus Alternaria are ubiquitously present and frequently found on fruits, vegetables and cereals. One of the most commonly found species from this genus is A. alternata which is able to produce the mycotoxin alternariol among others. To
A Solhaug et al.
Mutation research, 738-739, 1-11 (2012-10-04)
Alternariol (AOH), a mycotoxin produced by Alternaria sp, is often found as a contaminant in fruit and cereal products. Here we employed the murine macrophage cell line RAW 264.7 to test the hypothesis that AOH causes toxicity as a response

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service