Skip to Content
Merck
All Photos(1)

Key Documents

34342

Supelco

Fenbutatin oxide

PESTANAL®, analytical standard

Synonym(s):

Bis[tris-(2-methyl-2-phenylpropyl)tin] oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H78OSn2
CAS Number:
Molecular Weight:
1052.68
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)(C[Sn](CC(C)(C)c1ccccc1)(CC(C)(C)c2ccccc2)O[Sn](CC(C)(C)c3ccccc3)(CC(C)(C)c4ccccc4)CC(C)(C)c5ccccc5)c6ccccc6

InChI

1S/6C10H13.O.2Sn/c6*1-10(2,3)9-7-5-4-6-8-9;;;/h6*4-8H,1H2,2-3H3;;;

InChI key

HOXINJBQVZWYGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K A Barnes et al.
Rapid communications in mass spectrometry : RCM, 11(2), 159-164 (1997-01-01)
A rapid method, using high-performance liquid chromatography (HPLC)/ atmospheric pressure chemical ionization-mass spectrometry, has been developed for the determination of fenbutatin oxide in tomatoes, cucumbers and bananas. Samples were homogenized with sodium carbonate and ethyl acetate, filtered through sodium sulphate
R Montes et al.
Analytical and bioanalytical chemistry, 395(8), 2601-2610 (2009-10-07)
The fate of the acaricide fenbutatin oxide (FBTO) during the elaboration of white wine is evaluated. Matrix solid-phase dispersion (MSPD) and solid-phase microextraction (SPME) were used as sample preparation techniques applied to the semi-solid and the liquid matrices involved in
Adenir V Teodoro et al.
Experimental & applied acarology, 47(4), 293-299 (2008-11-15)
We used a Y-tube olfactometer to assess the sub-lethal effects of the acaricide fenbutatin oxide on the olfactory response of the predatory mite Iphiseiodes zuluagai towards odours from: (1) air or undamaged coffee plants; (2) undamaged or red spider mite
Fenbutatin oxide and chlorfenvinphos effects on the entomophagous arthropod fauna of citrus.
C Inglesfield
Bulletin of environmental contamination and toxicology, 38(5), 813-819 (1987-05-01)
Adenir V Teodoro et al.
Experimental & applied acarology, 36(1-2), 61-70 (2005-08-09)
Lethal concentration (LC) has been widely used to estimate pesticide toxicity. However, it does not consider the sub-lethal effects. Therefore we included the instantaneous rate of increase in association with LC to estimate population-level effects of the acaricides fenbutatin oxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service