Skip to Content
Merck
All Photos(1)

Key Documents

34223

Supelco

Azaperone

VETRANAL®, analytical standard

Synonym(s):

1-(4-Fluorophenyl)-4-(4-(2-pyridinyl)-1-piperazinyl)-1-butanone, 4′-Fluoro-4-[4-(2-pyridyl)-1-piperazinyl]butyrophenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22FN3O
CAS Number:
Molecular Weight:
327.40
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Fc1ccc(cc1)C(=O)CCCN2CCN(CC2)c3ccccn3

InChI

1S/C19H22FN3O/c20-17-8-6-16(7-9-17)18(24)4-3-11-22-12-14-23(15-13-22)19-5-1-2-10-21-19/h1-2,5-10H,3-4,11-15H2

InChI key

XTKDAFGWCDAMPY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Azaperone (AZN) is a butyrophenone neuroleptic drug, used as a tranquilizer in veterinary medicine.

Application

Azaperone may be used as an analytical reference standard for the determination of the analyte in animal kidneys and blood meal by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mari L Heinonen et al.
Veterinary anaesthesia and analgesia, 36(2), 151-157 (2009-02-26)
To investigate a combination of azaperone, detomidine, butorphanol and ketamine (DBK) in pigs and to compare it with the combination of azaperone, tiletamine and zolazepam (TZ). Prospective, randomized, blinded, cross-over study. Twelve clinically healthy crossbred pigs aged about 2 months
Determination of sedatives and adrenergic blockers in blood meal using accelerated solvent extraction and Orbitrap mass spectrometry.
Choi JH, et al.
Journal of Chromatography A, 1260(1-2), 111-119 (2012)
Lisa L Wolfe et al.
Journal of wildlife diseases, 44(3), 748-752 (2008-08-12)
Sixteen captive and five free-ranging black bears (Ursus americanus) were immobilized with a combination of butorphanol, azaperone, and medetomidine (BAM). The BAM drug combination was premixed using 0.5 ml butorphanol (30 mg/ml), 0.25 ml azaperone (50 mg/ml), and 0.25 ml
K Ruediger et al.
Journal of animal science, 90(7), 2331-2336 (2012-01-31)
This study examined the impact of a single dose of azaperone administered to sows at the end of farrowing on piglet weight gain and mortality during the lactation period. Two hundred fifty-two sows (JSR hybrid) housed in a conventional farrowing
Ramona D Donald et al.
Physiology & behavior, 104(5), 906-913 (2011-06-15)
The ability to assess emotionality is important within animal welfare research. Yet, for farm animals, few tests of emotionality have been well validated. Here we investigated the construct validity of behavioural measures of pig emotionality in an open-field test by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service