Skip to Content
Merck
All Photos(2)

Documents

34217

Supelco

Ronidazole-d3

VETRANAL®, analytical standard

Synonym(s):

2-(Carbamoyloxymethyl)-1-methyl-d3-5-nitro-imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D3H5N4O4
CAS Number:
Molecular Weight:
203.17
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

mass shift

M+3

SMILES string

[2H]C([2H])([2H])n1c(COC(N)=O)ncc1[N+]([O-])=O

InChI

1S/C6H8N4O4/c1-9-4(3-14-6(7)11)8-2-5(9)10(12)13/h2H,3H2,1H3,(H2,7,11)/i1D3

InChI key

PQFRTXSWDXZRRS-FIBGUPNXSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Ronidazole-d3 may be used internal standard in the determination of avermectins, benzimidazoles and nitroimidazoles residues in bovine muscle tissue using ultra-high performance liquid chromatography coupled with mass spectrometry (UHPLC/MS-MS).

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Salomé Martínez-Morcillo et al.
International journal of hygiene and environmental health, 223(1), 10-21 (2019-11-11)
This study assessed the presence of 27 pharmaceutically active compounds belonging to common therapeutic groups (cardiovascular, antiashmatic, psychoactive, diuretic, analgesic/anti-inflammatory, and antibiotic drugs) in the tissues of representative seafood species of bivalves, cephalopods, arthropods, and fish of high economic importance
Jèssica Subirats et al.
Molecular ecology, 26(20), 5567-5581 (2017-08-10)
Wastewater discharges introduce antibiotic residues and antibiotic-resistant bacteria (ARB) into surface waters. Both inputs directly affect the streambed resistome, either by exerting a selective pressure that favour the proliferation of resistant phenotypes or by enriching the resident communities with wastewater-associated
Belinda Huerta et al.
The Science of the total environment, 634, 542-549 (2018-04-11)
Fish are good indicators of aquatic environment pollution because of their capability to uptake pollutants contained in water. Therefore, accumulation of pharmaceutical compounds in freshwater and marine fish and other aquatic organisms has been studied extensively in the last decade.
Multiresidue method for identification and quantification of avermectins, benzimidazoles and nitroimidazoles residues in bovine muscle tissue by ultra-high performance liquid chromatography tandem mass spectrometry (UHPLC-MS/MS) using a QuEChERS approach
Silva DRG, et al.
Talanta, 171, 307-320 (2017)
Sara Rodriguez-Mozaz et al.
Environment international, 140, 105733-105733 (2020-05-01)
A comprehensive monitoring of a broad set of antibiotics in the final effluent of wastewater treatment plants (WWTPs) of 7 European countries (Portugal, Spain, Ireland, Cyprus, Germany, Finland, and Norway) was carried out in two consecutive years (2015 and 2016).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service