Skip to Content
Merck
All Photos(2)

Key Documents

34170

Supelco

Imidacloprid-d4

PESTANAL®, analytical standard

Synonym(s):

Imidazolidin-4,4,5,5-d4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9D4H6ClN5O2
CAS Number:
Molecular Weight:
259.69
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

mass shift

M+4

SMILES string

[2H]C1([2H])N=C(N[N+]([O-])=O)N(Cc2ccc(Cl)nc2)C1([2H])[2H]

InChI

1S/C9H10ClN5O2/c10-8-2-1-7(5-12-8)6-14-4-3-11-9(14)13-15(16)17/h1-2,5H,3-4,6H2,(H,11,13)/i3D2,4D2

InChI key

YWTYJOPNNQFBPC-KHORGVISSA-N

Related Categories

General description

Imidacloprid is a neonicotinoide insecticide primarily applied as a seed dressing and foliar sprays to protect crop in agricultural systems and landscaping.

Find more information here: Neonicotinoids

Application

Imidacloprid-d4 may be used as a surrogate standard in the determination of imidacloprid in honey and pollen samples using solid phase extraction liquid chromatography−tandem mass spectrometry method (SPE-LC-MS/MS).
Imidacloprid-d4 may be used as an internal standard for the determination of the analyte in soil and food matrices by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a fast and sensitive method for measuring multiple neonicotinoid insecticide residues in soil and the application in parks and residential areas.
Zhou Y, et al.
Analytica Chimica Acta, 1016, 19-28 (2018)
Determination of acetamiprid, imidacloprid, and spirotetramat and their relevant metabolites in pistachio using modified QuEChERS combined with liquid chromatography-tandem mass spectrometry.
Faraji M, et al.
Food Chemistry, 240, 634-641 (2018)
Ian Laycock et al.
PloS one, 8(11), e79872-e79872 (2013-11-14)
Currently, there is concern about declining bee populations and some blame the residues of neonicotinoid pesticides in the nectar and pollen of treated crops. Bumble bees are important wild pollinators that are widely exposed to dietary neonicotinoids by foraging in
Michelle L Hladik et al.
Environmental pollution (Barking, Essex : 1987), 193, 189-196 (2014-07-22)
Neonicotinoid insecticides are of environmental concern, but little is known about their occurrence in surface water. An area of intense corn and soybean production in the Midwestern United States was chosen to study this issue because of the high agricultural
Vijayakumar Bommuraj et al.
Food additives & contaminants. Part B, Surveillance, 13(4), 233-243 (2020-05-15)
Throughout the world, more than six billion people consume milk and milk products yearly. The safety and quality of dairy milk are regularly monitored in most countries worldwide. The Israeli monitoring program of chemical residues in milk has not changed

Articles

Analysis of 20 pharmaceutical compounds in calf serum using Supel™ Swift HLB SPE cartridge for cleanup and LC-MS determination and another commercially available HLB cartridge for comparison.

Analysis of 20 pharmaceutical compounds in calf serum using Supel™ Swift HLB SPE cartridge for cleanup and LC-MS determination and another commercially available HLB cartridge for comparison.

Protocols

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service