Skip to Content
Merck
All Photos(1)

Key Documents

34137

Supelco

Diacetoxyscirpenol solution

~100 μg/mL in acetonitrile, analytical standard

Synonym(s):

12,13-Epoxytrichothec-9-ene-3,4,15-triol-4,15-diacetate solution, 4β,15-Diacetoxy-3α-hydroxy-12,13 epoxy-trichothec-9-ene solution, Anguidin solution, DAS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26O7
CAS Number:
Molecular Weight:
366.41
EC Number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

~100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

CC(=O)OC[C@]12CCC(C)=C[C@H]1O[C@@H]3[C@H](O)[C@@H](OC(C)=O)[C@@]2(C)[C@]34CO4

InChI

1S/C19H26O7/c1-10-5-6-18(8-23-11(2)20)13(7-10)26-16-14(22)15(25-12(3)21)17(18,4)19(16)9-24-19/h7,13-16,22H,5-6,8-9H2,1-4H3/t13-,14-,15-,16-,17-,18-,19-/m1/s1

InChI key

AUGQEEXBDZWUJY-ZLJUKNTDSA-N

General description

Certan Vial

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

purity : ≥95.5% (HPLC)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yukio Yonemoto et al.
International journal of rheumatic diseases, 20(6), 702-707 (2016-03-19)
The objective of this study was to assess arthritis of the whole body before and after total knee arthroplasty (TKA) in patients with rheumatoid arthritis (RA) using positron emission tomography (PET). Seventeen knees of 17 RA patients (median age 68
Gabriella Santangelo et al.
Amyotrophic lateral sclerosis & frontotemporal degeneration, 18(5-6), 434-442 (2017-04-23)
Apathy is associated with cognitive decline and worse survival in amyotrophic lateral sclerosis (ALS); an accurate evaluation of this aspect is relevant in clinical settings. The aims of this study are to evaluate the prevalence of apathy in a large
Sytske Anne Bergstra et al.
Rheumatology (Oxford, England), 56(10), 1721-1728 (2017-09-29)
To compare 10-year disease outcomes of RA patients who have continuous low disease activity and are on MTX with or without initial combination therapy with infliximab or prednisone and SSZ. Recent-onset RA patients from the Behandel Strategieen (BeSt) (Dutch acronym
Gaoqiang Fei et al.
Traffic injury prevention, 20(6), 570-574 (2019-06-22)
Objective: Driving anger is a common emotion while driving and has been associated with traffic crashes. This study aimed to investigate situations that increase driving anger among Chinese drivers. Methods: A cross-sectional study was conducted among 3,101 drivers in southern
Chiara Pennetta et al.
Nanomaterials (Basel, Switzerland), 10(6) (2020-06-04)
A recently reported functionalization of single and multi-walled carbon nanotubes, based on a cycloaddition reaction between carbon nanotubes and a pyrrole derived compound, was exploited for the formation of a doxorubicin (DOX) stacked drug delivery system. The obtained supramolecular nano-conveyors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service