Skip to Content
Merck
All Photos(1)

Key Documents

34057

Supelco

Diclazuril

VETRANAL®, analytical standard

Synonym(s):

2,6-Dichloro-α-(4-chlorophenyl)-4-[4,5-dihydro-3,5-dioxo-1,2,4-triazin-2(3H)-yl]benzeneacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H9Cl3N4O2
CAS Number:
Molecular Weight:
407.64
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Clc1ccc(cc1)C(C#N)c2c(Cl)cc(cc2Cl)N3N=CC(=O)NC3=O

InChI

1S/C17H9Cl3N4O2/c18-10-3-1-9(2-4-10)12(7-21)16-13(19)5-11(6-14(16)20)24-17(26)23-15(25)8-22-24/h1-6,8,12H,(H,23,25,26)

InChI key

ZSZFUDFOPOMEET-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2

Target Organs

Lungs,Liver,lymph node

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assessing the effect of diclazuril on the intestinal absorptive capacity of broilers infected with experimental coccidiosis, using d-xylose absorption test.
H Nodeh et al.
Journal of veterinary pharmacology and therapeutics, 31(3), 265-267 (2008-05-13)
Xiaojiong Shen et al.
Veterinary parasitology, 185(2-4), 290-295 (2011-11-15)
Eimeria tenella (E. tenella) is one of the most virulent pathogens of coccidiosis. In apicomplexan parasites, Hsp90 (Heat shock protein 90) is essential for the invasion and survival in host cells. In this study, the effect of diclazuril, an effective
Zuhao Shi et al.
Se pu = Chinese journal of chromatography, 27(3), 303-307 (2009-10-07)
The effects of four pretreatment methods (acetonitrile extraction-evaporation concentration, acetonitrile extraction-solid phase extraction (SPE), matrix solid-phase dispersion (MSPD) extraction and MSPD-SPE) for the simultaneous analysis of diclazuril and toltrazuril residues in chicken tissues were compared. The average recovery of 70%
C Le Sueur et al.
Parasitology research, 104(5), 1157-1162 (2008-12-17)
A field study was conducted to evaluate the effect of a single oral treatment with 20 mg/kg body weight (BW) of toltrazuril (Baycox 5% suspension)--TOL--in comparison to a single oral treatment with 1 mg/kg BW of diclazuril (Vecoxan suspension orale
Bian-hua Zhou et al.
Parasitology research, 112(4), 1771-1780 (2013-02-19)
Screening the anticoccidial drug targets is very important for developing novel drugs and revealing the molecular basis of drug resistance in coccidia. Due to high effectivity and safety, diclazuril was used widely in the poultry industry. To assess the roles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service