Skip to Content
Merck
All Photos(2)

Key Documents

34056

Supelco

Propisochlor

PESTANAL®, analytical standard

Synonym(s):

2-Chloro-2′-ethyl-6′-methyl-N-(isopropoxymethyl)acetanilide, Proponit

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22ClNO2
CAS Number:
Molecular Weight:
283.79
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCc1cccc(C)c1N(COC(C)C)C(=O)CCl

InChI

1S/C15H22ClNO2/c1-5-13-8-6-7-12(4)15(13)17(14(18)9-16)10-19-11(2)3/h6-8,11H,5,9-10H2,1-4H3

InChI key

KZNDFYDURHAESM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Dermal

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhonghou Xu et al.
Journal of colloid and interface science, 287(2), 422-427 (2005-06-01)
The competitive sorption behavior of Cu2+ and propisochlor on humic acids (HA) was investigated using a batch method. The sorption equilibrium time of propisochlor on HA was 12 h when its initial concentration was 4 or 10 mg L(-1). The
Gang Xu et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 42(2), 165-171 (2007-03-17)
The effects of two humic acids (HAs) of different origins on the photodegradation of the chloroacetanilide herbicides acetochlor, propisochlor and butachlor were investigated in this study. One of the tested HAs was a standard sample that was purchased from a
Renáta Rauch et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 47(7), 670-676 (2012-05-09)
The catalytic effect of an oil shale rock "alginite" was studied under laboratory conditions on the decomposition of herbicide propisochlor. The breakdown process was followed in four parallel experiments: buffered solution (pH = 7), in buffer solutions containing alginite or
Yu Wang et al.
Huan jing ke xue= Huanjing kexue, 25(4), 94-99 (2004-11-02)
The synergetic effect of dissolved oxygen (DO) in the photodegradation of propisochlor aqueous solution was investigated in the presence of UV. With the increase of dissolved oxygen concentration, from 0 to 7.5 mg/L, photodegradation of propisochlor was accelerated. After the
G Wágner et al.
Communications in agricultural and applied biological sciences, 71(3 Pt A), 809-813 (2007-03-30)
Green pea (Pisum sativum L.), is one of the important vegetable crop in Hungary. Chemical weed control has an important role in pea growing. Pre-emergence herbicides are used most frequently in a green pea culture because they eliminate competition between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service