Skip to Content
Merck
All Photos(1)

Documents

34001

Supelco

Ceftiofur

VETRANAL®, analytical standard

Synonym(s):

3-[(2-Furylcarbonyl)thio]methyl]-7-[2-(2-amino-4-thiazolyl)-2-(methoxyiminoacetamido)-3-cephem-4-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17N5O7S3
CAS Number:
Molecular Weight:
523.56
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CO\N=C(/C(=O)N[C@H]1C2SCC(CSC(=O)c3ccco3)=C(N2C1=O)C(O)=O)c4csc(N)n4

InChI

1S/C19H17N5O7S3/c1-30-23-11(9-7-34-19(20)21-9)14(25)22-12-15(26)24-13(17(27)28)8(5-32-16(12)24)6-33-18(29)10-3-2-4-31-10/h2-4,7,12,16H,5-6H2,1H3,(H2,20,21)(H,22,25)(H,27,28)/b23-11-/t12-,16?/m1/s1

InChI key

ZBHXIWJRIFEVQY-FITYNHDMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ceftiofur is a third-generation cephalosporin antibiotic, which is commonly used in veterinary medicine for the treatment of respiratory diseases and bacterial infections in farm animals.

Application

Ceftiofur has been used as an analytical reference standard for the determination of the analyte in:
  • Milk samples using receptor binding assays such as Betastar Combo, SNAP, and Twinsensor tests.
  • Environmental water samples using capillary electrophoresis.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tate B Morris et al.
Frontiers in veterinary science, 7, 284-284 (2020-06-26)
The nociceptive blockade of locoregional anesthesia prior to surgical stimulation can decrease anesthetic agent requirement and thereby potential dose-dependent side effects. The use of an ipsilateral second and third cervical spinal nerve locoregional anesthetic block for prosthetic laryngoplasty in the
Sanjie Caol et al.
BMC veterinary research, 13(1), 293-293 (2017-10-01)
Lyme disease in humans is predominantly treated with tetracycline, macrolides or beta lactam antibiotics that have low minimum inhibitory concentrations (MIC) against Borrelia burgdorferi. Horses with Lyme disease may require long-term treatment making frequent intravenous or intramuscular treatment difficult and
Validation of receptor-binding assays to detect antibiotics in goat's milk.
Beltran CM, et al.
Journal of Food Protection, 77(2), 308-313 (2014)
Sam Abraham et al.
The ISME journal, 12(10), 2352-2362 (2018-06-15)
This study investigated the ecology, epidemiology and plasmid characteristics of extended-spectrum cephalosporin (ESC)-resistant E. coli in healthy pigs over a period of 4 years (2013-2016) following the withdrawal of ESCs. High carriage rates of ESC-resistant E. coli were demonstrated in
Céline Ster et al.
Veterinary research, 48(1), 56-56 (2017-09-25)
Staphylococcus aureus intramammary infections (IMIs) have low cure rates using standard antibiotic treatment and increasing the duration of treatment usually improves therapeutic success. Chronic IMIs are thought to be caused by bacteria presenting a specific virulence phenotype that includes the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service