Skip to Content
Merck

33366

Leptophos

PESTANAL®, analytical standard

Synonym(s):

O-(4-Bromo-2,5-dichlorophenyl) O-methyl phenylphosphonothioate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H10BrCl2O2PS
CAS Number:
Molecular Weight:
412.07
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
244-472-8
Beilstein/REAXYS Number:
2152663
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, NMR: suitable, gas chromatography (GC): suitable

mp

71-73 °C

suitability

passes test for identity (NMR)

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COP(=S)(Oc1cc(Cl)c(Br)cc1Cl)c2ccccc2

InChI

1S/C13H10BrCl2O2PS/c1-17-19(20,9-5-3-2-4-6-9)18-13-8-11(15)10(14)7-12(13)16/h2-8H,1H3

InChI key

CVRALZAYCYJELZ-UHFFFAOYSA-N

General description

Leptophos, an organophosphorus insecticide found to be persistent in the environment, shows neurotoxic effects in humans. Its mode of action for pest control involves the inhibition of acetylcholinesterase enzyme activity.

Application

Leptophos may be used as an analytical reference standard for the quantification of the analyte in vegetables using solid phase micro-extraction coupled with gas chromatography-flame photometric detector (SPME-GC-FPD).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Leptophos


signalword

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - STOT SE 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges

flash_point_f

>212.0 °F

flash_point_c

> 100 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library