Skip to Content
Merck
All Photos(2)

Key Documents

32952

Supelco

TCPP, mixture of isomers

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18Cl3O4P
CAS Number:
Molecular Weight:
327.57
Beilstein:
1842347
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

neat

SMILES string

CC(CCl)OP(=O)(OC(C)CCl)OC(C)CCl

InChI

1S/C9H18Cl3O4P/c1-7(4-10)14-17(13,15-8(2)5-11)16-9(3)6-12/h7-9H,4-6H2,1-3H3

InChI key

KVMPUXDNESXNOH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

TCPP, mixture of isomers is suitable for use in environmental and food residue analysis. It is a mixture of isomers, composition may vary, typical composition: main isomer tris(1-chloro-2-propyl) phosphate 66%, minor components: bis(1-chloro-2-propyl) (2-chloropropyl) phosphate and (1-chloro-2-propyl) bis(2-chloropropyl) phosphate.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

365.0 °F - closed cup

Flash Point(C)

185 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Regnery et al.
Journal of environmental monitoring : JEM, 13(2), 347-354 (2010-12-07)
Occurrence and distribution of chlorinated and non-chlorinated organophosphates in 72 groundwater samples from Germany under different recharge/infiltration conditions were investigated. Tris(2-chloro-1-methylethyl) phosphate (TCPP) and tris(2-chloroethyl) phosphate (TCEP) were the most frequently detected organophosphates in groundwater samples. Highest individual organophosphate concentrations
Lars Jurzik et al.
International journal of hygiene and environmental health, 213(3), 210-216 (2010-06-18)
There are still conflicting results on the suitability of chemical and microbiological parameters as indicators for the viral contamination of surface waters. In this study, conducted over 20 months, the abundance of human adenovirus, human polyomavirus, enterovirus, group A rotavirus
Julia Regnery et al.
Chemosphere, 78(8), 958-964 (2010-01-16)
To investigate seasonal fluctuations and trends of organophosphate (flame retardants, plasticizers) concentrations in rain and snow, precipitation samples were collected in 2007-2009 period at a densely populated urban sampling site and two sparsely populated rural sampling sites in middle Germany.
C van Netten et al.
American Industrial Hygiene Association journal, 52(9), 398-400 (1991-09-01)
Air sampling in a government building was necessary in response to reports of a cancer cluster. SKC (Eighty Four, Pa.) charcoal coconut shell-based sorbent tubes (226-01 lot 120) were recommended for this procedure. A recently purchased supply was present at
Na Li et al.
Ecotoxicology and environmental safety, 183, 109502-109502 (2019-08-09)
The urine levels of organophosphate flame retardants (PFRs) and bisphenol A (BPA) in kindergarten children (n = 31, 4-6 years old, sampling performed in 2016) in Hong Kong were measured. The detection frequency of the target PFRs, tri(2-chloroethyl)phosphate (TCEP), tris(1,3-dichloro-2-propyl) phosphate (TDCIPP)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service