Skip to Content
Merck
All Photos(1)

Key Documents

32205-M

Sigma-Aldrich

Ethyl alcohol, pure

≥99.8% (GC), suitable for UV/Vis spectroscopy, absolute, reag. Ph. Eur., USP/NF, reag. ISO

Synonym(s):

Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
EC Number:
MDL number:
UNSPSC Code:
15101802
PubChem Substance ID:
NACRES:
NA.21
technique(s):
UV/Vis spectroscopy: suitable
bp:
78 °C (lit.)
vapor pressure:
44.6 mmHg ( 20 °C)

Product Name

Ethanol, absolute, reag. ISO, reag. Ph. Eur., ≥99.8% (GC), liquid (clear, colorless)

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

Quality Level

vapor density

1.59 (vs air)

vapor pressure

44.6 mmHg ( 20 °C)

grade

absolute

Assay

≥99.8% (GC)

form

liquid (clear, colorless)

autoignition temp.

683 °F

expl. lim.

19 %, 60 °F

technique(s)

UV/Vis spectroscopy: suitable

impurities

furfural, in accordance
volatile impurities (GC), complies
≤0.0001% free alkali (as NH3)
≤0.0001% heavy metals (as Pb)
≤0.0003% KMnO4 red. matter (as O)
≤0.0005% free acid (as CH3COOH)
≤0.001% acetone (GC)
≤0.001% aldehydes (as CH3CHO)
≤0.001% non-volatile matter
≤0.003% 2-propanol
≤0.003% carbonyl compounds (as CO)
≤0.02% methanol (GC)
≤0.05% iso-amyl alcohol
≤0.2% water (Karl Fischer)

color

APHA: >10

refractive index

n20/D 1.3600 (lit.)
n20/D 1.3612-1.3618

pH

7 (20 °C, 10 g/L)

bp

78 °C (lit.)

mp

−114 °C (lit.)

cation traces

Al: ≤0.00005%
B: ≤0.000002%
Ba: ≤0.00001%
Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%

SMILES string

CCO

suitability

corresponds for UV absorption
passes test for reaction against H2SO4

format

neat

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethanol was used as dispersion medium for already prepared silver nanoparticles.
It may be used in the following studies:
  • Hydrothermal synthesis of single crystal ZnO nanorod.
  • Synthesis of N-(N′dodecylcarboxamidomethyl)N,N,dimethyl ammonio propane sulfonates.
  • As solvent for cytofluorimetric analysis.
  • Preparation of TiO2 colloid (particle size = 20nm).

Caution

Pertains only in Germany: Nicht zu Fertigungszwecken1

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesca Filon Larese et al.
Toxicology, 255(1-2), 33-37 (2008-11-01)
There is a growing interest on nanoparticle safety for topical use. The benefits of nanoparticles have been shown in several scientific fields, but little is known about their potential to penetrate the skin. This study aims at evaluating in vitro
Christophe Tastet et al.
Proteomics, 3(2), 111-121 (2003-02-26)
Several zwitterionic detergents differing in their polar heads, linker parts and hydrophobia tail were synthesized and evaluated for their efficiency in protein solubilizers for two-dimensional electrophoresis. A model system consisting of human red blood cell ghosts was used for this
Fabrication of thin film dye sensitized solar cells with solar to electric power conversion efficiency over 10%.
Ito S, et al.
Thin Solid Films, 516(14), 4613-4619 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service