Skip to Content
Merck
All Photos(1)

Key Documents

32096

Supelco

Indaziflam

PESTANAL®, analytical standard

Synonym(s):

2-Amino-4-[(1R,2S)-2,6-dimethyl-indan-1-ylamino]-6-(1-fluoroethyl)-1,3,5-triazine, N-[(1R,2S)-2,3-Dihydro-2,6-dimethyl-1H-inden-1-yl]-6-[(1RS)-1-fluoroethyl]-1,3,5-triazine-2,4-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20FN5
CAS Number:
Molecular Weight:
301.36
Beilstein:
20920435
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(F)c1nc(N)nc(N[C@@H]2[C@@H](C)Cc3ccc(C)cc23)n1

InChI

1S/C16H20FN5/c1-8-4-5-11-7-9(2)13(12(11)6-8)19-16-21-14(10(3)17)20-15(18)22-16/h4-6,9-10,13H,7H2,1-3H3,(H3,18,19,20,21,22)/t9-,10?,13+/m0/s1

InChI key

YFONKFDEZLYQDH-BOURZNODSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Indaziflam is a pre-emergent, alkylazine herbicide, used in weed control management. Its mode of action involves inhibiting cellulose wall synthesis in susceptible plants.

Application

Indaziflam may be used as a reference standard in the determination of indaziflam and its metabolite residues in grapes using liquid-liquid extraction with high performance liquid chromatography coupled with ultraviolet detection (HPLC-UV).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Indaziflam, a new herbicide for pre-emergent control of weeds in turf, forestry, industrial vegetation and ornamentals
Kaapro Jand Hall J
Pakistan journal of weed science research, 18(1), 267-270 (2012)
Determination of Indaziflam and its metabolite residues in black grapes
Revathi P, et al.
International Journal of Chemical Studies, 5(2), 237-241 (2017)
PRE and POST control of annual bluegrass (Poa annua) with indaziflam
Brosnan.TJ, et al.
Weed Technology, 26(1), 48-53 (2012)
Smooth crabgrass control with indaziflam at various spring timings
Brosnan.TJ, et al.
Weed Technology, 25(3), 363-366 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service