Skip to Content
Merck
All Photos(1)

Key Documents

32056

Supelco

Carbofuran

PESTANAL®, analytical standard

Synonym(s):

2,3-Dihydro-2,2-dimethyl-7-benzofuranol N-methylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO3
CAS Number:
Molecular Weight:
221.25
Beilstein:
1428746
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

150-153 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

CNC(=O)Oc1cccc2CC(C)(C)Oc12

InChI

1S/C12H15NO3/c1-12(2)7-8-5-4-6-9(10(8)16-12)15-11(14)13-3/h4-6H,7H2,1-3H3,(H,13,14)

InChI key

DUEPRVBVGDRKAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Carbofuran belongs to N-methylcarbamate class of chemicals used to control soil-dwelling and foliar-feeding insects on agricultural crops. Carbofuran acts by inhibiting cholinesterase enzyme activity.[1]

Application

Carbofuran may be used as a reference standard for the determination of the analyte:
  • In well water samples by high-performance liquid chromatography employing a variable-wavelength ultraviolet detector set.[2]
  • In fruits and vegetables by monoclonal enzyme immunoassay and high-performance liquid chromatography with fluorescence detection.[3]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Development of monoclonal antibody-based immunoassays to the N-methylcarbamate pesticide carbofuran.
Abad A, et al.
Journal of Agricultural and Food Chemistry, 47(6), 2475- 2485 (1999)
Determination of carbaryl, carbofuran and methiocarb in cucumbers and strawberries by monoclonal enzyme immunoassays and high-performance liquid chromatography with fluorescence detection: An analytical comparison.
Abad A, et al.
Journal of Chromatography A, 833(1), 3-12 (1999)
Determination of aldicarb, aldicarb sulfoxide, aldicarb sulfone and carbofuran residues in water using high-performance liquid chromatography.
Cochrane WP, et al.
Journal of Chromatography A, 243(2), 307- 314 (1982)
Ahmed S A Harabawy et al.
Ecotoxicology and environmental safety, 103, 61-67 (2014-01-28)
The present work aimed to evaluate the toxic effects of two sublethal concentrations of carbofuran pesticide (0.16 and 0.49mg/L, for 35 days) on hematological and blood biochemical parameters of catfish, Clarias gariepinus, and to evaluate the genotoxic potential of carbofuran
Ratnesh Kumar Sharma et al.
Disease markers, 32(3), 153-163 (2012-03-02)
Various efforts have been made in past in order to predict the underlying mechanism of pesticide-induced toxicity using in vitro and animal models, however, these predictions may or may not be directly correlated with humans. The present study was designed

Protocols

-methylcarbamate 10 μg/mL; Diuron; Propham; Siduron; Methiocarb, analytical standard; Linuron 10 μg/mL; Swep 10 μg/mL; Chlorpropham 10 μg/mL; Barban; Neburon

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service