Skip to Content
Merck
All Photos(1)

Documents

31527

Supelco

Terbumeton

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19N5O
CAS Number:
Molecular Weight:
225.29
Beilstein:
520400
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCNc1nc(NC(C)(C)C)nc(OC)n1

InChI

1S/C10H19N5O/c1-6-11-7-12-8(15-10(2,3)4)14-9(13-7)16-5/h6H2,1-5H3,(H2,11,12,13,14,15)

InChI key

BCQMBFHBDZVHKU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Terbumeton is a selective and systemic, s-triazine herbicide widely used for weed control management.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Terbumeton may be used as a reference standard in the determination of terbumeton and its metabolites in soil bulk water using solid phase extraction with high-performance liquid chromatography coupled with diode array detector (HPLC-DAD).

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junwei Liu et al.
The Science of the total environment, 579, 115-123 (2016-11-22)
s-Triazine herbicides have been widely used in recent decades and caused serious concern over contamination of groundwater, surface water and soil. A novel bacterial strain JW-1 was isolated from activated sludge and identified as Leucobacter sp. based on comparative morphology
Surface engineering: molecularly imprinted affinity membranes by photograft polymerization
Advanced Environmental and Chemical Sensing Technology, 4205, 65-75 (2001)
Analysis of terbumeton and its major metabolites by SPE and DAD-HPLC in soil bulk water
Conrad A, et al.
Chromatographia, 65(3-4), 155-161 (2007)
Yue Li et al.
Journal of chromatography. A, 1567, 81-89 (2018-07-23)
A novel method for the determination of sixteen polycyclic aromatic hydrocarbons (PAHs) in environmental water samples was developed using column clean-up (CCU) coupled with continuous flow single drop microextraction (CF-SDME) prior to the determination by gas chromatography-mass spectrometry (GC-MS). In
Xinpei Li et al.
Mikrochimica acta, 185(7), 336-336 (2018-06-25)
This work describes a nanohybrid material consisting of gold nanoparticles (AuNPs) and nanosheets of layered double hydroxides (NLDHs) of Mg(II) and Al(III). Mono-disperse AuNPs were immobilized on the surface of the LDHs via Au-O bonding. The nanohybrid sorbent was packed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service