Skip to Content
Merck
All Photos(1)

Key Documents

31014

Supelco

Mercury(II) sulfate

ACS reagent, for preparation of solution for COD determination according to DIN 38409, part 41, ≥99%

Synonym(s):

Mercuric sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HgSO4
CAS Number:
Molecular Weight:
296.65
EC Number:
MDL number:
UNSPSC Code:
41113319
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
for preparation of solution for COD determination according to DIN 38409, part 41

Quality Level

Assay

≥99%

analyte functional class(es)

chemical oxygen demand (COD)

technique(s)

photometry: suitable

ign. residue (after red. with HCOOH)

≤0.02% (as SO4)

anion traces

chloride (Cl-): ≤30 mg/kg
nitrate (NO3-): ≤30 mg/kg

cation traces

Ca: ≤30 mg/kg
Cd: ≤1 mg/kg
Cu: ≤5 mg/kg
Fe: ≤10 mg/kg
Hg+: ≤500 mg/kg
K: ≤20 mg/kg
Mg: ≤30 mg/kg
Na: ≤50 mg/kg
Pb: ≤5 mg/kg

application(s)

environmental
general analytical

SMILES string

[Hg++].[O-]S([O-])(=O)=O

InChI

1S/Hg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

DOBUSJIVSSJEDA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ai-hua Liang et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 30(4), 249-252 (2005-02-24)
Traditional Chinese medicine (TCM) experienced a gradual course in recognition of the toxicity of Cinnabaris from "nontoxic" to "toxic". The ancient doctors of TCM understood both the toxic property and the regularity of increasing toxicity of Cinnabaris. In long-term clinical
Chao-jun Chen et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 37(19), 2968-2970 (2012-12-29)
Herein, the synthesis, component, microstructure and pharmacological and toxicology researches of the Synthetic Mercury Sulfide (S-HgS) a kind of common drug in Chinese, Mongolia, Tibetan medicine, and Indian medicine system were summarized. The similar cognition about mercury toxicity & pharmacological
[A case of granulomatous reaction to tattooing].
R Colombo et al.
Minerva chirurgica, 42(8), 747-749 (1987-04-30)
[Morphological manifestations of the toxic effect of uni- and bivalent mercury compounds during repeated peroral administration].
A P Samoĭlov et al.
Vrachebnoe delo, (9)(9), 107-109 (1983-09-01)
B Mora et al.
Canadian journal of microbiology, 26(8), 930-937 (1980-08-01)
Studies have been made on the toxicities of three inorganic (HgCl2, HgSO4, and NH2HgCl) and two organic (CH3HgCl and phenyl mercuric acetate (PMA)) mercury derivatives on planktonic algae (Nitzschia acicularis W. Sm. and Tetraselmis suecica Butch.) Growth kinetics and flourescence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service