Skip to Content
Merck
All Photos(1)

Key Documents

27715

Sigma-Aldrich

L-(+)-Lactic acid

80%

Synonym(s):

(S)-2-Hydroxypropionic acid, Sarcolactic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O3
CAS Number:
Molecular Weight:
90.08
Beilstein:
1720251
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

Assay

80%

form

liquid

ign. residue

≤0.1% (as SO4)

solubility

water: soluble(lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Ca: ≤100 mg/kg
Fe: ≤10 mg/kg

antibiotic activity spectrum

Gram-negative bacteria

Mode of action

cell membrane | interferes

SMILES string

C[C@H](O)C(O)=O

InChI

1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)/t2-/m0/s1

InChI key

JVTAAEKCZFNVCJ-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Pharmaceutical and biomaterial applications: L-(+)-Lactic acid enhances M2 macrophage polarization through regulation of the PCK2/AMPK/mTOR signaling pathway in poly-L-lactic acid membranes, indicating its vital role in biomedicine and materials science (Tang et al., 2024).

  • Agricultural biotechnology: The induction of defense responses in tobacco against Phytophthora nicotianae by L-(+)-Lactic acid underscores its potential application in agricultural biotechnology for plant disease management (Yan et al., 2024).



Biochem/physiol Actions

Use as a substrate for the lactic acid dehydrogenase enzymes.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis and characterization of pH-sensitive hydrogels based on chitosan and D, L-lactic acid.
Qu X, et al.
Journal of Applied Polymer Science, 74(13), 3193-3202 (1999)
Carlos Carmona-Fontaine et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(11), 2934-2939 (2017-03-02)
The genetic and phenotypic diversity of cells within tumors is a major obstacle for cancer treatment. Because of the stochastic nature of genetic alterations, this intratumoral heterogeneity is often viewed as chaotic. Here we show that the altered metabolism of
Recovery of L-(+)-lactic acid by anion exchange resin Amberlite IRA-400.
Cao X, et al.
Biochemical Engineering Journal, 11(2), 189-196 (2002)
Swelling and hydrolytic degradation of poly (D, L-lactic acid) in aqueous solutions.
Proikakis C S, et al.
Polymer Degradation and Stability, 91(3), 614-619 (2006)
T Isotalo et al.
BJU international, 88(1), 30-34 (2001-07-12)
To assess whether patients in acute urinary retention from benign prostatic enlargement can be treated with a combined therapy comprising finasteride and a bioabsorbable self-reinforced poly L-lactic acid (SR-PLLA) urethral stent. Fifty-five patients in acute urinary retention were treated as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service