Skip to Content
Merck
All Photos(1)

Key Documents

274348

Sigma-Aldrich

TWEEN® 20

average Mn ~1,228

Synonym(s):

Polyethylene glycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900

description

non-ionic

form

viscous liquid

mol wt

~1228
average Mn ~1,228

technique(s)

ELISA: suitable
PCR: suitable
blocking: suitable
cell culture | mammalian: suitable
immunohistochemistry: suitable
protein purification: suitable

refractive index

n20/D 1.468 (lit.)

density

1.095 g/mL at 25 °C (lit.)

HLB

16

SMILES string

CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO

InChI

1S/C26H50O10/c1-2-3-4-5-6-7-8-9-10-11-24(30)34-19-18-31-20-22(32-15-12-27)26-25(35-17-14-29)23(21-36-26)33-16-13-28/h22-23,25-29H,2-21H2,1H3

InChI key

HMFKFHLTUCJZJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Non-ionic detergent

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhou Yu et al.
Scientific reports, 8(1), 9972-9972 (2018-07-04)
The relationship between autophagy and tumour is well studied, but tumour cell morphological changes associated with autophagy defects are rarely reported, especially in renal cell carcinoma (RCC). We collected 10 renal tumour samples with characteristic eosinophilic cytoplasmic inclusions (ECIs) and
Aida Maddahi et al.
BMC neuroscience, 9, 85-85 (2008-09-17)
MEK1/2 is a serine/threonine protein that phosphorylates extracellular signal-regulated kinase (ERK1/2). Cerebral ischemia results in enhanced expression of cerebrovascular contractile receptors in the middle cerebral artery (MCA) leading to the ischemic region. Here we explored the role of the MEK/ERK
Sang Jun Han et al.
Genesis (New York, N.Y. : 2000), 47(12), 815-824 (2009-11-03)
Estrogen receptor alpha (ERalpha), plays essential roles in the female reproduction. To investigate the dynamic changes in ERalpha activity in vivo, we have developed an ER Alpha Activity Indicator (ERAAI) mouse. This ERAAI mouse harbors both a modified ERalpha Bacterial
Junxia Tang et al.
Human molecular genetics, 18(20), 3851-3863 (2009-07-21)
DTNBP1 (dystrobrevin binding protein 1) remains a top candidate gene in schizophrenia. Reduced expression of this gene and of its encoded protein, dysbindin-1, have been reported in the brains of schizophrenia cases. It has not been established, however, if the
Sergiy Kostenko et al.
Cellular and molecular life sciences : CMLS, 68(5), 847-862 (2010-08-25)
The mitogen-activated protein kinase-activated protein kinase-5 (MK5) resides predominantly in the nucleus of resting cells, but p38(MAPK), extracellular signal-regulated kinases-3 and -4 (ERK3 and ERK4), and protein kinase A (PKA) induce nucleocytoplasmic redistribution of MK5. The mechanism by which PKA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service