251283
Sulfanilic acid sodium salt hydrate
97%
Synonym(s):
4-aminobenzenesulfonic acid sodium salt hydrate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
4-(H2N)C6H4SO3Na·xH2O
CAS Number:
Molecular Weight:
195.17 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Assay:
97%
form:
crystals
Recommended Products
Quality Level
Assay
97%
form
crystals
functional group
sulfonic acid
SMILES string
O.[Na+].Nc1ccc(cc1)S([O-])(=O)=O
InChI
1S/C6H7NO3S.Na.H2O/c7-5-1-3-6(4-2-5)11(8,9)10;;/h1-4H,7H2,(H,8,9,10);;1H2/q;+1;/p-1
InChI key
VDGKZGAQOPUDQL-UHFFFAOYSA-M
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Xiaomin Zhu et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 10(16), 3871-3878 (2004-08-19)
The synthesis of 4-N-[3',4',5'-tris(dodecyloxy)benzamido]benzene-4-sulfonic acid (1) and 4'-[3",4",5"-tris(dodecyloxy)benzoyloxy]azobenzene-4-sulfonic acid (2) is described. Pure acid 1 is stable, while 2 can be stored only in solution. Both acids were obtained from their sodium salts and were quantitatively transformed into the pyridinium
Synthesis, chemical modification, and surface assembly of carbon nanowires.
Amma A, et al.
Advances in Functional Materials, 13(5), 365-370 (2003)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service