Skip to Content
Merck
All Photos(2)

Key Documents

244813

Sigma-Aldrich

Reactive Blue 4

Dye content 35 %

Synonym(s):

1-Amino-4-[3-(4,6-dichlorotriazin-2-ylamino)-4-sulfophenylamino]anthraquinone-2-sulfonic acid, Procion® blue MX-R

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H14Cl2N6O8S2
CAS Number:
Molecular Weight:
637.43
Colour Index Number:
61205
Beilstein:
602258
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

composition

Dye content, 35%

λmax

595 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Nc1c(cc(Nc2ccc(c(Nc3nc(Cl)nc(Cl)n3)c2)S(O)(=O)=O)c4C(=O)c5ccccc5C(=O)c14)S(O)(=O)=O

InChI

1S/C23H14Cl2N6O8S2/c24-21-29-22(25)31-23(30-21)28-12-7-9(5-6-14(12)40(34,35)36)27-13-8-15(41(37,38)39)18(26)17-16(13)19(32)10-3-1-2-4-11(10)20(17)33/h1-8,27H,26H2,(H,34,35,36)(H,37,38,39)(H,28,29,30,31)

InChI key

RTLULCVBFCRQKI-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Reactive blue 4 (C.l. 61205) is a dichlorotriazine dye also known as procion brilliant blue MX-R, procion brilliant blue M.

Application

Reactive blue dyes have been used for labeling antibodies. Such anthraquinone dyes are widely used as immobilized ligands in systems for protein separations. Reactive blue 4 is used as a tracer to assess apical leakage in dental fillings. It is routinely used to stain proteins on electrophoretic gels.

Legal Information

Procion is a registered trademark of Dystar Textilfarben GmbH & Co.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gülay Bayramoğlu et al.
Journal of hazardous materials, 137(3), 1689-1697 (2006-06-13)
The native and treated fungal biomass of Phanerocheate chrysosporium was used for the biosorption of a textile dye (i.e., Reactive Blue 4). In the batch system, the biosorption equilibrium time was about 4 h and the maximum dye uptake on
Belgin Gözmen et al.
Journal of hazardous materials, 168(1), 129-136 (2009-03-10)
The degradations of an anthraquinone dye, the reactive blue 4 (RB4), were studied by wet air oxidation (WAO), wet peroxide oxidation (WPO), photocatalytic oxidation, and electro-Fenton (EF) advanced oxidation. The RB4 oxidation was evaluated by the decrease in total organic
Shih-Hsien Chang et al.
Journal of hazardous materials, 166(2-3), 1127-1133 (2009-01-17)
Degradation of two different kinds of dyes, anthraquinone Reactive Blue 4 (RB4) and azo Reactive Black 5 (RB5), by low-cost zero valent iron (Fe(0)) in a N(2) bubbling system (Fe(0)/N(2) process) and air bubbling system (Fe(0)/air process) was investigated. The
Characterization of the textile anthraquinone dye Reactive Blue 4
Epolito W J
Dyes and Pigments, 67, 35-46 (2005)
Young H Lee et al.
Water environment research : a research publication of the Water Environment Federation, 78(2), 156-169 (2006-03-29)
The decolorization of two anthraquinone dyes (Reactive Blue 4 [RB4] and Reactive Blue 19 [RB19]) and two phthalocyanine dyes (Reactive Blue 7 [RB7] and Reactive Blue 21 [RB21]) was investigated at an initial dye concentration of 300 mg/L using an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service