Skip to Content
Merck
All Photos(1)

Documents

23454

Supelco

Bisphenol M

analytical standard

Synonym(s):

4,4′-(1,3-Phenylenediisopropylidene)bisphenol, Bisphenol M

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4[C(CH3)2C6H4OH]2
CAS Number:
Molecular Weight:
346.46
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

135-139 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CC(C)(c1ccc(O)cc1)c2cccc(c2)C(C)(C)c3ccc(O)cc3

InChI

1S/C24H26O2/c1-23(2,17-8-12-21(25)13-9-17)19-6-5-7-20(16-19)24(3,4)18-10-14-22(26)15-11-18/h5-16,25-26H,1-4H3

InChI key

PVFQHGDIOXNKIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bisphenol M is a bisphenol analog, used in a variety of industrial applications, such as the synthesis of polycarbonate and epoxy resins. It is also used in electronics, toys, food containers, bottles, papers, medical equipment, etc.

Application

Bisphenol M may be used as an analytical reference standard for the quantification of the analyte in human breast milk and beverages using gas chromatography coupled with tandem mass spectrometry (GC-MS/MS). It may also be used as an analytical standard for the green chemical synthesis of phosphorus containing polymers via gas-liquid or solid-liquid interfacial polycondensation with phenylphosphonic dichloride.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bisphenol analogues other than BPA: environmental occurrence, human exposure, and toxicity? a review
Chen D, et al.
Environmental Science & Technology, 50(11), 5438-5453 (2016)
Biodegradation of bisphenol compounds in the surface water of Taihu Lake and the effect of humic acids
Zhou N et al.
The Science of the Total Environment (2020)
A dispersive liquid-liquid microextraction based on solidification of floating organic droplet followed by injector port silylation coupled with gas chromatography?tandem mass spectrometry for the determination of nine bisphenols in bottled carbonated beverages
Mandrah K, et al.
Journal of Chromatography A, 1528(1), 10-17 (2017)
Green synthesis of polymers containing phosphorus in the main chain
Iliescu S, et al.
Comptes Rendus Chimie, 14(7-8), 647-651 (2011)
V Petruševski et al.
Journal of pharmaceutical and biomedical analysis, 124, 228-235 (2016-03-12)
The chemical characterization of plastics for pharmaceutical packaging has been subject to ever increasing regulatory scrutiny, the reasons for which being: a) plastic additives and degradation products can be extremely hazardous to the patients' health (especially patients on chronic therapy)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service