Skip to Content
Merck
All Photos(1)

Key Documents

23310

Sigma-Aldrich

2-Chloroaniline

technical, ≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NH2
CAS Number:
Molecular Weight:
127.57
Beilstein:
606077
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.588

bp

208-210 °C (lit.)

solubility

water: soluble 5.13 g/L at 20 °C

density

1.213 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

Nc1ccccc1Cl

InChI

1S/C6H6ClN/c7-5-3-1-2-4-6(5)8/h1-4H,8H2

InChI key

AKCRQHGQIJBRMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

  • 2-Chloroaniline is widely used in polymer, rubber, pharmaceutical and dye industries.
  • Determination of 2-chloroaniline in polymer industrial wastewater has been examined using HPLC with on-line microdialysis.
  • Degradation of 2-chloroaniline in water by ozonation as well as by photolysis (UV-light of 254 nm) and radiolysis (γ-rays) in the presence of ozone has been investigated.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Muta. 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative studies on the degradation of aqueous 2-chloroaniline by O3 as well as by UV-light and ?-rays in the presence of ozone.
Winarno EK and Getoff N.
Radiation Physics and Chemistry, 65(4), 387-395 (2002)
H Kataoka
Journal of chromatography. A, 733(1-2), 19-34 (1996-05-10)
The environmental analysis of amines is important to preserve human health because these compounds often have toxic effects. Gas chromatography (GC) of free amines is generally unsatisfactory owing to the adsorption and decomposition of the solutes on the column. Derivatization
Dmitry Tsvelikhovsky et al.
Journal of the American Chemical Society, 132(40), 14048-14051 (2010-09-23)
The Pd-catalyzed condensation of 2-bromostyrene and 2-chloroaniline derivatives yields stable diphenylamine intermediates, which are selectively converted to five-, six-, or seven-membered heteroaromatics (indoles, carbazoles, acridines, and dibenzazepines). The selectivity of these intramolecular transformations is uniquely ligand-controlled and offers efficient routes
Salvatore Andini et al.
Journal of hazardous materials, 157(2-3), 599-604 (2008-02-22)
Fuel oil fly ash has been tested as low-cost carbon-based adsorbent of 2-chlorophenol (CP), 2-chloroaniline (CA) and methylene blue (MB) from aqueous solutions. In all the cases the adsorption was found to be of cooperative type. Different adsorption capacities were
L Zampori et al.
Journal of hazardous materials, 170(2-3), 1041-1049 (2009-06-16)
The aim of the present work is to investigate the effect of a prolonged leaching test (more than a year) on the microstructure of solidified cementitious wasteforms. A set of four different cement-based monoliths (Ap, Bp, Cp and Dp) was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service