Key Documents
20998
Hamilton® syringe, 1000 series GASTIGHT®, PTFE luer lock
1002TLL, PTFE Luer lock, volume 2.5 mL, needle size (not included)
About This Item
UNSPSC Code:
41122001
eCl@ss:
32070500
Pricing and availability is not currently available.
material
PTFE Luer lock
needle size
(not included)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81420
needle L
, n/a
volume
2.5 mL
fitting
luer lock (without slots)
compatibility
for use with Hamilton 1002TLL (PTFE Luer lock)
General description
Hamilton syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed.[1] The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.[2]
Application
It has been used during calorimetric analysis for phthalate protonation in different ionic media.[3]
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Critical evaluation of protonation constants. Literature analysis and experimental potentiometric and calorimetric data for the thermodynamics of phthalate protonation in different ionic media.
Bretti, Clemente, et al.
Journal of Solution Chemistry, 35.9, 1227-1244 (2006)
Guy, Jeffrey S.
Pharmacology for the Prehospital Professional, 54-54 (2009)
Wolf, Don P.
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 06-Dec, 192-192 (2012)