Key Documents
20708
Hamilton® GASTIGHT® syringe, 1700 series
1750LTN, volume 0.5 mL, needle size 22 ga (side-port), needle L 51 mm (2 in.)
About This Item
UNSPSC Code:
41122001
Pricing and availability is not currently available.
material
PTFE plunger tip
cone tip, side port stainless steel needle (point style 5)
needle size
22 ga (side-port)
description
Gastight®
feature
needle type N (cemented)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81243
needle L
51 mm (2 in.)
volume
0.5 mL
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)