Skip to Content
Merck
All Photos(3)

Documents

198161

Sigma-Aldrich

Toluidine Blue O

certified by the Biological Stain Commission

Synonym(s):

Basic Blue 17, Blutene chloride, Methylene Blue T50 or T extra, Tolonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16ClN3S
CAS Number:
Molecular Weight:
305.83
Colour Index Number:
52040
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

certified by the Biological Stain Commission

Quality Level

form

powder

composition

Dye content, 80%

color

dark green

λmax

626 nm

ε (extinction coefficient)

≥13000 at 241-247 nm in H2O
≥28000 at 287-293 nm in H2O
≥40000 at 625-635 nm in H2O

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].Cc1cc2N=C3C=CC(\C=C3Sc2cc1N)=[N+](\C)C

InChI

1S/C15H15N3S.ClH/c1-9-6-13-15(8-11(9)16)19-14-7-10(18(2)3)4-5-12(14)17-13;/h4-8,16H,1-3H3;1H

InChI key

GEDVVYWLPUPJJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A metachromatic dye suitable for a wide variety of histological staining procedures.

Application

Toluidine Blue O along with light-emitting diode has been used as a photosensitizer to control in vitro biofilms and to kill gram-negative bacteria.

Biochem/physiol Actions

Toluidine Blue O (TBO) is a thiazine dye of the quinone-imine family and is cationic in nature. It has been employed in polychromatic staining of paraffin embedded plant cell walls. TBO can be removed from its aqueous solution by using an effective adsorbent, clinoptilolite. TBO and fixed output laboratory laser light source are used together to destroy variety of oral bacteria associated with dental caries. This metachromatic nuclear stain is often used in place of thionin or methylene blue. It also serves as a component of a tribasic stain along with Malachite Greenand Basic Fuchsin.

Suitability

Certified for use as a metachromatic stain for mast cells and cartilage using the Churukian-Schenk method and the method of Nocht on paraffin sections.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photosensitization of in vitro biofilms by toluidine blue O combined with a light-emitting diode.
Zanin IC
European Journal of Oral Sciences, 114(1), 64-69 (2006)
Factors influencing the susceptibility of Gram-negative bacteria to toluidine blue O-mediated lethal photosensitization
komerik N
Journal of Applied Microbiology, 92(4), 618?623-618?623 (2002)
The adsorption kinetics and removal of cationic dye, Toluidine Blue O, from aqueous solution with Turkish zeolite
Alpat S K
Journal of Hazardous Materials, 151(1), 213-220 (2008)
A TRIBASIC STAIN FOR THIN SECTIONS OF PLASTIC-EMBEDDED, OSO4-FIXED TISSUES.
P M GRIMLEY
Stain technology, 39, 229-233 (1964-07-01)
The Effect of Variable Energy Input from a Novel Light Source on the Photoactivated Bactericidal Action of Toluidine Blue O on Streptococcus mutans
Williams J A
Caries Research, 37, 190?193-190?193 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service