Skip to Content
Merck
All Photos(1)

Key Documents

15236

Sigma-Aldrich

N,O-Bis(trimethylsilyl)carbamate

≥98.0% (T)

Synonym(s):

BSC, Trimethylsilyl N-(trimethylsilyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiNHCO2Si(CH3)3
CAS Number:
Molecular Weight:
205.40
Beilstein:
2043399
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (T)

form

solid

mp

77-83 °C

functional group

amine

SMILES string

C[Si](C)(C)NC(=O)O[Si](C)(C)C

InChI

1S/C7H19NO2Si2/c1-11(2,3)8-7(9)10-12(4,5)6/h1-6H3,(H,8,9)

InChI key

DGIJAZGPLFOQJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Extremely suitable reagent for the silylation of alcohols, phenols and carboxylic acids. The only by-products are CO2 and NH3[1]; Silyloxycarbonylation of amines[2][3]; Acylisocyanates from carboxylic acid chlorides[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ronak Saluja et al.
Journal of oncology practice, 14(5), e280-e294 (2018-03-31)
The purpose of this study was to determine if clinical benefits of novel anticancer drugs, measured by the ASCO Value Framework and European Society of Medical Oncology (ESMO) Magnitude of Clinical Benefit Scale, have increased over time in parallel with
Yi-Long Wu et al.
BMC cancer, 11, 430-430 (2011-10-11)
Previous research suggests the therapeutic cancer vaccine L-BLP25 potentially provides a survival benefit in patients with locally advanced unresectable stage III non-small cell lung carcinoma (NSCLC). These promising findings prompted the phase III study, INSPIRE, in patients of East-Asian ethnicity.
Gregor Vlacic et al.
Cells, 8(9) (2019-09-19)
Malignant pleural mesothelioma (MPM) is a devastating malignancy with limited therapeutic options. Fibroblast growth factor receptors (FGFR) and their ligands were shown to contribute to MPM aggressiveness and it was suggested that subgroups of MPM patients could benefit from FGFR-targeted
L. Birkofer et al.
Journal of Organometallic Chemistry, 99, C1 (1975)
Sudarshan Chari et al.
PLoS genetics, 9(8), e1003661-e1003661 (2013-08-13)
The phenotypic outcome of a mutation cannot be simply mapped onto the underlying DNA variant. Instead, the phenotype is a function of the allele, the genetic background in which it occurs and the environment where the mutational effects are expressed.

Articles

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service