Skip to Content
Merck
All Photos(1)

Key Documents

14407

Sigma-Aldrich

N-(+)-Biotinyl-6-aminohexanoic acid

≥97.0% (HPLC)

Synonym(s):

(+)-Biotin-ε-aminocaproic acid, 6-((Biotinoyl)amino)hexanoic acid, Biotin-X, N-(+)-Biotinyl-6-aminocaproic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H27N3O4S
CAS Number:
Molecular Weight:
357.47
Beilstein:
706392
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.79

Quality Level

Assay

≥97.0% (HPLC)

form

powder

color

colorless to white

mp

220-230 °C (dec.)

cation traces

Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤100 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

detection

SMILES string

[H][C@]12CS[C@@H](CCCCC(=O)NCCCCCC(O)=O)[C@@]1([H])NC(=O)N2

InChI

1S/C16H27N3O4S/c20-13(17-9-5-1-2-8-14(21)22)7-4-3-6-12-15-11(10-24-12)18-16(23)19-15/h11-12,15H,1-10H2,(H,17,20)(H,21,22)(H2,18,19,23)/t11-,12-,15-/m0/s1

InChI key

CMUGHZFPFWNUQT-HUBLWGQQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Biotinylation reagent
N-(+)-Biotinyl-6-aminohexanoic acid is suitable to perform biotinylation.

Biochem/physiol Actions

Biotinyl-6-aminohexanoic acid (Biotin-X) may be used in the biosynthesis of biotinylated oligosaccharides. Biotinyl-6-aminohexanoic acid is frequently used as a derivative of N-hydroxysuccinimide ester.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert P Picone et al.
Molecular pharmacology, 68(6), 1623-1635 (2005-09-15)
The CB1 cannabinoid receptor has been shown to play important physiological roles in the central nervous system, as well as peripherally, and is a target for development of therapeutic medications. To gain insight on the ligand binding site(s) and structural
Multivalent foldamer-based affinity assay for selective recognition of A beta oligomers
Olajos G, et al.
Analytica Chimica Acta, 960, 131-137 (2017)
C R Valeri et al.
Vox sanguinis, 88(2), 122-129 (2005-02-22)
In accordance with Food and Drug Administration (FDA) regulations, platelets can be stored in the liquid state at 22 degrees C for only 5 days. Platelets frozen with 6% dimethylsulphoxide (DMSO) can be stored at -80 degrees C for 2
In vivo labeling of Escherichia coli cell envelope proteins with N-hydroxysuccinimide esters of biotin.
Bradburne JA, Godfrey P, et al.
Applied and Environmental Microbiology, 59, 663-668 (2993)
Infrared analysis of biomolecule attachment to functionalized silicon surfaces
Biointerface Characterization by Advanced IR Spectroscopy, 83-113 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service