Skip to Content
Merck
All Photos(3)

Key Documents

131164

Sigma-Aldrich

Indigo carmine

certified by the Biological Stain Commission, certified by the BSC, Dye content 85 %

Synonym(s):

Acid Blue 74, Indigo-5,5′-disulfonic acid disodium salt, Indigocarmine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H8N2Na2O8S2
CAS Number:
Molecular Weight:
466.35
Colour Index Number:
73015
Beilstein:
4103904
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

certified by the Biological Stain Commission

Quality Level

Agency

certified by the BSC

Assay

>85.0% (HPLC)

form

solid

composition

Dye content, 85%

color

dark blue to very dark blue

pH

3.87 (30 °C, 7.14 g/L)

mp

≥340 °C

density

0.71 g/cm3 at 29 °C

λmax

608 nm

εmax

20,000 at 250-256 nm in water at 0.01 g/L
29500 at 284-290 nm in water at 0.01 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[O-]S(=O)(=O)c1ccc2NC(\C(=O)c2c1)=C3\Nc4ccc(cc4C3=O)S([O-])(=O)=O

InChI

1S/C16H10N2O8S2.2Na/c19-15-9-5-7(27(21,22)23)1-3-11(9)17-13(15)14-16(20)10-6-8(28(24,25)26)2-4-12(10)18-14;;/h1-6,17-18H,(H,21,22,23)(H,24,25,26);;/q;2*+1/p-2/b14-13+;;

InChI key

KHLVKKOJDHCJMG-QDBORUFSSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Indigo carmine has been used to investigate its effects on ozone production by neutrophils.

Biochem/physiol Actions

Indigo Carmine (IC), commonly known as acid blue 74 or food blue 1 or FD and C blue 2 is a blue synthetic dye. It is used as a redox indicator in analytical chemistry and as a microscopic stain in biology. IC is also employed as a photometric detector. IC is a highly toxic indigoid dye and is associated with various health hazards such as irritation to the gastrointestinal tract leading to nausea, vomiting, and diarrhea and irritation in the respiratory tract leading to coughing and shortness of breath. IC exposure might also cause skin and eye irritations to humans. IC dye spray is used for the diagnosis of dysplasia in ulcerative colitis and is considered to be a simple, feasible, and safe method.

Suitability

Used as a collagen stain and in procedures using red nuclear stains.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Superoxide converts indigo carmine to isatin sulfonic acid: implications for the hypothesis that neutrophils produce ozone.
Kettle AJ
The Journal of Biological Chemistry, 279(18), 18521-18525 (2004)
An experimental study of indigo carmine removal from aqueous solution by electrocoagulation
Secula M S
Desalination, 277(1-3), 227-235 (2011)
J. Ind. Chem. Soc., 63, 533-533 (1986)
Pancolonic indigo carmine dye spraying for the detection of dysplasia in ulcerative colitis
Rutter M D
Gut, 53(2), 256-260 (2004)
Rice husk ash as an effective adsorbent: evaluation of adsorptive characteristics for Indigo Carmine dye.
Lakshmi UR
Journal of Environmental Management, 90(2), 710-720 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service