Skip to Content
Merck
All Photos(2)

Key Documents

11344

Supelco

Disperse Yellow 3

analytical standard

Synonym(s):

4-(2-Hydroxy-5-methylphenylazo)acetanilide, N-[4-(2-Hydroxy-5-methylphenylazo)phenyl]acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15N3O2
CAS Number:
Molecular Weight:
269.30
Colour Index Number:
11855
Beilstein:
1915225
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥96.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

268-270 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CC(=O)Nc1ccc(cc1)N=Nc2cc(C)ccc2O

InChI

1S/C15H15N3O2/c1-10-3-8-15(20)14(9-10)18-17-13-6-4-12(5-7-13)16-11(2)19/h3-9,20H,1-2H3,(H,16,19)

InChI key

PXOZAFXVEWKXED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Disperse Yellow 3 is an yellow azo dye which is majorly used in textile industry. It also has carcinogenic properties.

Application

It was used to understand that Phanerochaete chrysosporium can mineralize azo dyes with radiolabeled substrate.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J T Spadaro et al.
Applied and environmental microbiology, 58(8), 2397-2401 (1992-08-01)
Under nitrogen-limiting, secondary metabolic conditions, the white rot basidiomycete Phanerochaete chrysosporium extensively mineralized the specifically 14C-ring-labeled azo dyes 4-phenylazophenol, 4-phenylazo-2-methoxyphenol, Disperse Yellow 3 [2-(4'-acetamidophenylazo)-4-methylphenol], 4-phenylazoaniline, N,N-dimethyl-4-phenylazoaniline, Disperse Orange 3 [4-(4'-nitrophenylazo)-aniline], and Solvent Yellow 14 (1-phenylazo-2-naphthol). Twelve days after addition to
B M Hausen et al.
Deutsche medizinische Wochenschrift (1946), 109(39), 1469-1475 (1984-09-28)
Skin allergies caused by the wearing of stockings and hose have received little attention. Findings in patients of an allergy department, enquiries at stocking counters of stores and recent publications indicate, however, that probably many more persons have an allergy
Disperse Yellow 3.
IARC monographs on the evaluation of carcinogenic risks to humans, 48, 149-159 (1990-01-01)
Case report: trouser dermatitis.
J H Apted
The Australasian journal of dermatology, 26(2), 80-80 (1985-08-01)
Varun Ahuja et al.
Archives of toxicology, 83(7), 691-699 (2009-02-13)
We used a modified protocol of the murine local lymph node assay (LLNA) to study the cross-sensitising potential of (a) textile dye disperse yellow 3 and its metabolite 2-amino-p-cresol, (b) two antibiotics, penicillin G and cefotiam. The test substances were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service