Skip to Content
Merck
All Photos(1)

Key Documents

09258

Supelco

Erythrodiol

analytical standard

Synonym(s):

Olean-12-ene-3β,28-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H50O2
CAS Number:
Molecular Weight:
442.72
Beilstein:
2340203
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
NACRES:
NA.21

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

230-231 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

[H][C@@]12CC(C)(C)CC[C@]1(CO)CC[C@]3(C)C2=CC[C@]4([H])[C@@]5(C)CC[C@H](O)C(C)(C)[C@]5([H])CC[C@@]34C

InChI

1S/C30H50O2/c1-25(2)14-16-30(19-31)17-15-28(6)20(21(30)18-25)8-9-23-27(5)12-11-24(32)26(3,4)22(27)10-13-29(23,28)7/h8,21-24,31-32H,9-19H2,1-7H3/t21-,22-,23+,24-,27-,28+,29+,30+/m0/s1

InChI key

PSZDOEIIIJFCFE-OSQDELBUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Erythrodiol is a natural triterpenoid produced in olives. It is believed to exhibit antiproliferative and proapoptotic activity in colon adenocarcinoma cells.

Application

Erythrodiol has been used as working standard for determination of terpenoid in olive leaves using SPE followed by HPLC analysis.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean Wandji et al.
Planta medica, 68(9), 822-826 (2002-10-03)
Chemical studies of the CH2 Cl2 -MeOH extract of the seeds of Gambeya africana (Baker) Pierre led to the isolation of 15 compounds. Their structures were established on the basis of spectroscopic methods and chemical reactions. They comprised five new
Lucía Olmo-García et al.
Food chemistry, 239, 631-639 (2017-09-07)
Pentacyclic triterpenes are minor, but very relevant compounds found in virgin olive oil (VOO). A rapid and reliable LC-MS method for determining the triterpenic acids and dialcohols (after ultrasound assisted extraction) from VOO has been developed, giving an alternative to
Dalila Smati et al.
Journal of ethnopharmacology, 95(2-3), 405-407 (2004-10-28)
3beta-(3,4-Dihydroxycinnamoyl)-erythrodiol was isolated as the cytotoxic constituent of the roots of Zygophyllum geslini.
Javier S Perona et al.
Journal of agricultural and food chemistry, 53(3), 730-735 (2005-02-03)
In contrast to other metabolic functions, the role of dietary antioxidants and oil on microsomal lipid oxidation has been less extensively studied. This study examines ascorbate-Fe(2+) and NADPH-induced lipid peroxidation of hepatic microsomes of rats that were fed for three
Ayumi Ohsaki et al.
Journal of natural products, 67(3), 469-471 (2004-03-27)
Four new triterpenoids with various skeletons, maytefolins A-C (1-3) and uvaol-3-caffeate (4), were isolated from the leaves of a Brazilian medicinal plant, Maytenus ilicifolia, together with five known triterpenoids. Of these triterpenoids only erythrodiol exhibited significant cytotoxicity against KB/S, KB/VJ300

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service