Skip to Content
Merck

Skip To

08751

4-Amino-3-hydroxy-1-naphthalenesulfonic acid

for spectrophotometric det. of Si, ≥90.0% (CHN)

Synonym(s):

1-Amino-2-naphthol-4-sulfonic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
H2NC10H5(OH)SO3H
CAS Number:
Molecular Weight:
239.25
UNSPSC Code:
41116105
NACRES:
NA.21
PubChem Substance ID:
EC Number:
204-147-3
Beilstein/REAXYS Number:
2697469
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

assay

≥90.0% (CHN)

form

powder

quality

for spectrophotometric det. of Si

technique(s)

UV/Vis spectroscopy: suitable

mp

290 °C (dec.) (lit.)

SMILES string

Nc1c(O)cc(c2ccccc12)S(O)(=O)=O

InChI

1S/C10H9NO4S/c11-10-7-4-2-1-3-6(7)9(5-8(10)12)16(13,14)15/h1-5,12H,11H2,(H,13,14,15)

InChI key

RXCMFQDTWCCLBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
39896970862157031
assay

≥90.0% (CHN)

assay

≥90%

assay

-

assay

98%

form

powder

form

powder

form

crystals

form

-

quality

for spectrophotometric det. of Si

quality

-

quality

-

quality

-

technique(s)

UV/Vis spectroscopy: suitable

technique(s)

-

technique(s)

-

technique(s)

-

mp

290 °C (dec.) (lit.)

mp

290 °C (dec.) (lit.)

mp

280 °C (dec.) (lit.)

mp

305 °C (dec.) (lit.)

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

General description

4-Amino-3-hydroxy-1-naphthalenesulfonic acid is an aromatic amino-sulfonic acid.

Application

4-Amino-3-hydroxy-1-naphthalenesulfonic acid has been used to study separation of aromatic sulfonated compounds using Ion-interaction high-performance liquid chromatography and micellar electrokinetic capillary chromatography techniques. It has also been used as a starting material which when reacted with palmitoyl chloride yields the following derivatives:
  • 2′-Pentadecylnaphth[3,4-d]oxazole-1-sulfonic acid
  • 4-Amino-3-(palmitoyloxy)-1-naphthalenesufonic acid
  • 4-(Palmitoylamino)-3-hydroxy-1-naphthalenesulfonic acid
  • 4-(Palmitoylamino)-3-(palmitoyloxy)-1-naphthalenesulfonic acid

Other Notes

Reagent for the spectrophotometric det. of silicon

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F.A. Sorrentino et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 15, 441-441 (1970)
Synthesis of naphthalenesulfonic acid small molecules as selective inhibitors of the DNA polymerase and ribonuclease H activities of HIV-1 reverse transcriptase.
Mohan P
Journal of Medicinal Chemistry, 37(16), 2513-2519 (1994)
Ion-interaction high-performance liquid chromatography and micellar electrokinetic capillary chromatography: two complementary techniques for the separation of aromatic sulfonated compounds.
Angelino, S., et al.
Journal of Chromatography A, 845.1, 257-271 (1999)

Global Trade Item Number

SKUGTIN
W267708-5KG-K04061837806988
1A00900-25MG04065269096726
1A02470-25MG04065269107453
821243025004022536476605
821243000004022536476582
59932-1ML04061832644738
W267708-10KG-K04061837806957
W267708-1KG-K04061837806964
W267708-25KG-K04061837806971
W267708-SAMPLE-K04061837515910
821243100004022536476612
821243999904022536476629
M26615-100G04061834045939

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service