Skip to Content
Merck
All Photos(2)

Key Documents

08393

Supelco

Oleamide

analytical standard

Synonym(s):

cis-9,10-Octadecenoamide, cis-9-Octadecenamide, Oleic acid amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H35NO
CAS Number:
Molecular Weight:
281.48
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (TLC)

shelf life

limited shelf life, expiry date on the label

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCC(N)=O

InChI

1S/C18H35NO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H2,19,20)/b10-9-

InChI key

FATBGEAMYMYZAF-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Biochem/physiol Actions

Sleep-inducing brain lipid, which allosterically modulates GABAA receptors and potentiates 5-HT7 serotonin receptor responses. Selective endogenous agonist of rat and human CB1 cannabinoid receptor.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

404.6 °F - closed cup

Flash Point(C)

207 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Christopher J Fowler
British journal of pharmacology, 141(2), 195-196 (2003-12-24)
The fatty acid amide class of compounds, which include the endocannabinoid anandamide and the sleep-inducing compound oleamide, have been shown in vitro to have a multiplicity of actions upon different neurochemical systems. In the present issue of this journal, Leggett
Gregory P Mueller et al.
Vitamins and hormones, 81, 55-78 (2009-08-04)
Oleamide (cis-9-octadecenamide) is the prototype long chain primary fatty acid amide lipid messenger. The natural occurrence of oleamide was first reported in human serum in 1989. Subsequently oleamide was shown to accumulate in the cerebrospinal fluid of sleep-deprived cats and
Hiroshi Nojima et al.
Current drug safety, 2(3), 204-211 (2008-08-12)
Despite considerable research, metastasis remains a major challenge in the clinical management of cancer. Recent reports show that abnormally augmented expression of Cx26 is responsible for the enhanced spontaneous metastasis of mouse BL6 melanoma cells. The function of Cx26 appears
W B Mendelson et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 25(5 Suppl), S36-S39 (2001-10-30)
Oleamide is an endogenous fatty acid amide which can be synthesized de novo in the mammalian nervous system, and has been detected in human plasma. It accumulates in the CSF of rats after six hours of sleep deprivation and induces
C J Fowler et al.
Biochemical pharmacology, 62(5), 517-526 (2001-10-05)
Fatty acid amide hydrolase (FAAH) is responsible for the hydrolysis of a number of important endogenous fatty acid amides, including the endogenous cannabimimetic agent anandamide (AEA), the sleep-inducing compound oleamide, and the putative anti-inflammatory agent palmitoylethanolamide (PEA). In recent years

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service