07982
Sodium chloride physiological solution
BioUltra, tablet
Synonym(s):
Physiological Saline, tablets, Saline, physiological, tablets
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
product line
BioUltra
Quality Level
form
tablet
SMILES string
[Na]Cl
InChI
1S/ClH.Na/h1H;/q;+1/p-1
InChI key
FAPWRFPIFSIZLT-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Application
Sodium chloride physiological solution has been used in a study that demonstrated the effects of goblet cell-targeting nanoparticles on the oral absorption of insulin in vitro, ex vivo and in vivo. Sodium chloride physiological solution has also been used in a study to assess the utility of the hydrophobically-modified alginate gel system in the repair of articular cartilage defects.
Packaging
100 tablets in plastic bottles
Reconstitution
1 tablet in 100 mL of water yields 0.9% NaCl
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Sang Ho Park et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 67(Pt 10), 1250-1252 (2011-11-22)
Transmembrane protein 16A (TMEM16A, also known as anoctamin 1; ANO1) is a bona fide Ca(2+)-activated chloride channel that is activated by intracellular Ca(2+)- and Ca(2+)-mobilizing stimuli and plays important roles in a variety of physiological functions. To elucidate the structural
Mohammad Kazem Ghahramanpoor et al.
Journal of materials science. Materials in medicine, 22(10), 2365-2375 (2011-08-13)
Alginate is a family of natural polysaccharides, widely used in industry and medicine for many applications, with its non toxic nature, gentle sol/gel transition procedure and low cost, alginate inferior biomechanical properties have limited its utility especially in tissue engineering.
Yun Jin et al.
Biomaterials, 33(5), 1573-1582 (2011-11-19)
The present study was to demonstrate the effects of goblet cell-targeting nanoparticles on the oral absorption of insulin in vitro, ex vivo and in vivo, and identify the targeting mechanism as well as the influence of mucus. The insulin loaded
Mustafa S Atta et al.
Oxidative medicine and cellular longevity, 2018, 7845681-7845681 (2018-12-05)
Diabetic cardiomyopathy is a diabetic complication due to oxidative stress injuries. This study examined the protecting influence of thymoquinone (TQ) on diabetes-caused cardiac complications. The intracellular means by which TQ works against diabetes-caused cardiac myopathy in rats is not completely
Zhenghong Xu et al.
Journal of medicinal chemistry, 54(21), 7567-7578 (2011-10-05)
Membrane-associated serine protease matriptase has been implicated in human diseases and might be a drug target. In the present study, a novel class of matriptase inhibitors targeting zymogen activation is developed by a combination of the screening of compound library
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service