Skip to Content
Merck
All Photos(1)

Key Documents

07596

Supelco

Sodium butyrate

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Butyric acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2COONa
CAS Number:
Molecular Weight:
110.09
Beilstein:
3629439
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage condition

under inert gas (Argon)

mp

250-253 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

[Na+].CCCC([O-])=O

InChI

1S/C4H8O2.Na/c1-2-3-4(5)6;/h2-3H2,1H3,(H,5,6);/q;+1/p-1

InChI key

MFBOGIVSZKQAPD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Application

Sodium butyrate is a sodium salt of butyric acid, with potent anticancer activity. It can act like a histone deacetylase inhibitor and it is known to influence the gene expression and differentiation in many cell lines.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium Butyrate Induces NIH3T3 cells to senescence-like state and enhances promoter activity of p21WAF/CIP1in p53-Independent manner.
Xiao H, et al.
Biochemical and Biophysical Research Communications, 237(2), 457-460 (1997)
Fight Cancer with Vitamins and Supplements: A Guide to Prevention and Treatment (2001)
Youngmi Cho et al.
Experimental biology and medicine (Maywood, N.J.), 239(3), 302-310 (2014-02-06)
DNA methylation and histone acetylation contribute to the transcriptional regulation of genes involved in apoptosis. We have demonstrated that docosahexaenoic acid (DHA, 22:6 n-3) and butyrate enhance colonocyte apoptosis. To determine if DHA and/or butyrate elevate apoptosis through epigenetic mechanisms thereby
Jing Jiang et al.
Scientific reports, 5, 9191-9191 (2015-03-18)
Epithelial barrier integrity is critical to maintain the homeostasis in the body. The regulatory mechanism of the epithelial barrier function has not been fully understood. This study aims to elucidate the role of the TWIK-related potassium channel-1 (Trek1) in the
Anastasia Gelgor et al.
Journal of virology, 89(10), 5298-5307 (2015-03-06)
The Kaposi's sarcoma-associated herpesvirus (KSHV) open reading frame 16 (orf16) encodes a viral Bcl-2 (vBcl-2) protein which shares sequence and functional homology with the Bcl-2 family. Like its cellular homologs, vBcl-2 protects various cell types from apoptosis and can also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service