Skip to Content
Merck
All Photos(1)

Key Documents

07205

Supelco

α-Tocotrienol

analytical standard

Synonym(s):

D-α-Tocotrienol, (R)-α-Tocotrienol, [R-(E,E)]-3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyl-3,7,11-tridecatrienyl)-2H-1-benzopyran-6-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H44O2
CAS Number:
Molecular Weight:
424.66
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

biological source

Elaeis guineensis

Quality Level

grade

analytical standard

Assay

≥97.0% (HPLC)

form

liquid
viscous liquid

shelf life

limited shelf life, expiry date on the label

color

colorless to amber

application(s)

detection
vitamins, nutraceuticals, and natural products

format

neat

storage temp.

−20°C

SMILES string

C\C(C)=C\CC\C(C)=C\CC\C(C)=C\CC[C@]1(C)CCc2c(C)c(O)c(C)c(C)c2O1

InChI

1S/C29H44O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h12,14,16,30H,9-11,13,15,17-19H2,1-8H3/b21-14+,22-16+/t29-/m1/s1

InChI key

RZFHLOLGZPDCHJ-XZXLULOTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Tocotrienol is one of the isomers of vitamin E.

Application

α-Tocotrienol may be used as a reference standard for the analysis of α-Tocotrienol in cereals, in chicken meat and various food samples by high performance liquid chromatography (HPLC) equipped with fluorescence detection and in hazelnuts by normal phase HPLC combined with diode array detector (DAD).

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of tocopherols and tocotrienols in hazelnuts by a normal phase liquid chromatographic method.
Amaral JS, et al.
Analytical Sciences, 21(12), 1545-1548 (2005)
Application of supercritical fluid chromatography in the quantitative analysis of minor components (carotenes, vitamin E, sterols, and squalene) from palm oil.
Choo YM, et al.
Lipids, 40(4), 429-432 (2005)
Method for simultaneous analysis of eight vitamin E isomers in various foods by high performance liquid chromatography and fluorescence detection.
Pinheiro-Sant?Ana HM, et al.
Journal of Chromatography A, 1218(47), 8496-8502 (2011)
A small-scale sample preparation method with HPLC analysis for determination of tocopherols and tocotrienols in cereals.
Ryynanen M, et al.
J. Food Compos. Anal., 17(6), 749-765 (2004)
Simultaneous determination of vitamin E homologs in chicken meat by liquid chromatography with fluorescence detection.
Hewavitharana AK, et al.
Journal of Chromatography A, 1025(2), 313-317 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service