Skip to Content
Merck
All Photos(1)

Key Documents

06689

Supelco

3-Methylglutaconic acid, mixture of E and Z isomers

≥98.0% (HPLC)

Synonym(s):

3-Methyl-2-pentenedioic acid, NSC 249232

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O4
CAS Number:
Molecular Weight:
144.13
Beilstein:
1722907
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Quality Level

Assay

≥98.0% (HPLC)

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

C/C(CC(O)=O)=C\C(O)=O

InChI

1S/C6H8O4/c1-4(2-5(7)8)3-6(9)10/h2H,3H2,1H3,(H,7,8)(H,9,10)/b4-2+

InChI key

WKRBKYFIJPGYQC-DUXPYHPUSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

3-Methylglutaconic acid is a metabolite (as the CoA thioester) in the leucine degradative pathway as well as the mevalonate shunt, a pathway that links isoprenoid metabolism with mitochondrial acetyl-CoA metabolism. 3-Methylglutaconic acid accumulates in patients with a deficiency of 3-methylglutaconyl-CoA hydratase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R I Kelley
Clinica chimica acta; international journal of clinical chemistry, 220(2), 157-164 (1993-11-15)
A method is described for quantification of the trace metabolite, 3-methylglutaconic acid, by isotope-dilution gas chromatography/mass spectrometry using synthetic 3-[2,4,6-13C3]methylglutaconic acid. Results are shown for quantification of 3-methylglutaconic acid in plasma, urine, cerebrospinal fluid and amniotic fluid for both normal
E Holme et al.
Pediatric research, 32(6), 731-735 (1992-12-01)
We report the finding of mitochondrial ATP-synthase deficiency in a child with persistent 3-methylglutaconic aciduria. The child presented in the neonatal period with severe lactic acidosis, which was controlled by Na-HCO3 and glucose infusions. During the 1st y of life
H Costeff et al.
Annals of neurology, 33(1), 103-104 (1993-01-01)
Behr's syndrome consists of recessively inherited infantile optic atrophy, together with chronic neurological disturbances such as ataxia, extrapyramidal dysfunction, and juvenile spastic paresis. This syndrome was found to be relatively common among Iraqi Jews. For our study, 18 such patients
G A Mitchell et al.
Prenatal diagnosis, 15(8), 725-729 (1995-08-01)
We report the first molecular prenatal diagnosis of 3-hydroxy-3-methylglutaryl CoA lyase (HL) deficiency. The proband had a classic but severe presentation with hypoketotic hypoglycaemia and acidosis, secondary mental retardation, and epilepsy, and HL deficiency was documented in cultured fibroblasts. We
R I Kelley et al.
Pediatric research, 37(5), 671-674 (1995-05-01)
The branched-chain organic acid, 3-methylglutaconic acid, is an intermediate (as the CoA thioester) in the leucine degradative pathway as well as the mevalonate shunt, a pathway that links isoprenoid metabolism with mitochondrial acetyl-CoA metabolism. Because the majority of patients with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service