Skip to Content
Merck
All Photos(1)

Key Documents

03641

Supelco

Dansyl chloride

for HPLC derivatization, LiChropur, ≥99.0% (HPLC)

Synonym(s):

5-(Dimethylamino)naphthalene-1-sulfonyl chloride, DNSCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12ClNO2S
CAS Number:
Molecular Weight:
269.75
Beilstein:
2217205
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

grade

derivatization grade ((HPLC))
for HPLC derivatization

Quality Level

Assay

≥99.0% (HPLC)

form

powder or crystals

quality

LiChropur

technique(s)

HPLC: suitable

mp

72-74 °C (lit.)

solubility

acetone: 0.2 g/10 mL, clear to slightly turbid, yellow to very dark yellow-orange

storage temp.

2-8°C

SMILES string

CN(C)c1cccc2c(cccc12)S(Cl)(=O)=O

InChI

1S/C12H12ClNO2S/c1-14(2)11-7-3-6-10-9(11)5-4-8-12(10)17(13,15)16/h3-8H,1-2H3

InChI key

XPDXVDYUQZHFPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dansyl chloride is a fluorogenic reagent which is typically employed for the derivatization of amino acids.

Application

A fluorogenic reagent for fluorescent N-terminal derivatization of amino acids and peptides and detection by reverse phase HPLC.
Dansyl chloride has been used for the fluorogenic labeling of xenoestrogens, 4-nonylphenol and bisphenol A prior to their determination by high-performance liquid chromatography (HPLC) with fluorescence detection. It has also been used for the derivatization of icariin in human serum samples after oral administration of Epimedium decoction, followed by its determination using ultrasensitive LC-tandem mass spectrometry (LC-MS) and for the derivatization of antitumoral agent ES-285 in dog plasma followed by its analysis by HPLC equipped with ultraviolet and fluorescence detection.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phenylisothiocyanate and dansyl chloride precolumn derivatizations for the high-performance liquid chromatography analysis of the antitumoral agent ES-285 in dog plasma.
Maraschiello, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 791(1-2), 1-11 (2003)
Trace analysis of icariin in human serum with dansyl chloride derivatization after oral administration of Epimedium decoction by liquid chromatography tandem mass spectrometry.
Gong, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 860(2), 166-172 (2007)
Tomofumi Santa
Biomedical chromatography : BMC, 25(1-2), 1-10 (2010-11-09)
Liquid chromatography/electrospray ionization tandem mass spectrometry (LC/ESI-MS/MS) is one of the most prominent analytical techniques owing to its inherent selectivity and sensitivity. In LC/ESI-MS/MS, chemical derivatization is often used to enhance the detection sensitivity. Derivatization improves the chromatographic separation, and
Determination of the xenoestrogens 4-nonylphenol and bisphenol A by high-performance liquid chromatography and fluorescence detection after derivatisation with dansyl chloride.
Naassner, et al.
Journal of Chromatography A, 945(1-2), 133-138 (2002)
Miguel López-Gómez et al.
Phytochemistry, 107, 32-41 (2014-09-16)
Polyamines (PAs) are low molecular weight aliphatic compounds that have been shown to be an important part of plant responses to salt stress. For that reason in this work we have investigated the involvement of PAs in the response to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service