Skip to Content
Merck

Skip To

03450

N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride

purum, ≥98.0% (AT)

Synonym(s):

N-Ethyl-N′-(3-dimethylaminopropyl)carbodiimide hydrochloride, EDAC, EDC, EDC hydrochloride, WSC hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C8H17N3 · HCl
CAS Number:
Molecular Weight:
191.70
UNSPSC Code:
12352111
NACRES:
NA.31
PubChem Substance ID:
EC Number:
247-361-2
Beilstein/REAXYS Number:
5764110
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

Quality Segment

assay

≥98.0% (AT)

form

powder

mp

110-115 °C

solubility

H2O: soluble 1 gm/10 ml, clear to very slightly hazy, colorless to very faintly yellow

application(s)

microbiology

storage temp.

−20°C

SMILES string

Cl.CCN=C=NCCCN(C)C

InChI

1S/C8H17N3.ClH/c1-4-9-8-10-6-5-7-11(2)3;/h4-7H2,1-3H3;1H

InChI key

FPQQSJJWHUJYPU-UHFFFAOYSA-N

General description

1-Ethyl-3-(3-dimethylaminopropyl) carbodiimide hydrochloride (EDAC HCl), commonly known as EDAC, is a crucial water-soluble reagent widely employed in chemical and biochemical research, particularly for its proficiency in mediating amide bond formation. Notably effective in peptide synthesis, EDAC HCl demonstrates efficiency in coupling amino acids through their carboxyl and amine groups, enabling the tailored creation of peptides with specific sequences and functionalities. Extending its utility, EDAC HCl plays a pivotal role in the construction of immunogens by covalently attaching haptens—small immune-response eliciting molecules—to carrier proteins, a critical aspect in vaccine research.

The adaptability of EDAC HCl encompasses nucleic acid modification, permitting the selective labeling of DNA and RNA through their 5′ phosphate groups. This capacity contributes significantly to the visualization, tracking, and analytical aspects of these fundamental molecules, thereby advancing nucleic acid research. Furthermore, EDAC HCl functions as a biomolecule bridge, acting as a crosslinker connecting amine-reactive NHS-esters of biomolecules to carboxyl groups.

This feature proves invaluable in protein conjugation, facilitating the development of hybrid molecules with distinct properties and functions. The underlying reaction mechanism involves EDAC HCl′s interaction with a carboxyl group, forming an unstable intermediate actively seeking an amine partner. The delicate equilibrium of this reaction underscores the necessity for optimizing conditions to ensure efficient conjugation. The assistance of N-hydroxysuccinimide (NHS) enhances the capabilities of EDAC HCl by stabilizing the intermediate and enabling two-step conjugation procedures, affording greater flexibility and control, particularly in the manipulation of complex biomolecular structures.

Application

N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride has been used modify the cell surface of Escherichia coli to covalently couple substances. It has also been used as an activator to modify microfluidic chips to capture Escherichia coli.

Biochem/physiol Actions

N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide is a water soluble condensing reagent. EDAC is generally utilized as a carboxyl activating agent for amide bonding with primary amines. Additionally, it reacts with phosphate groups. It has been utilized in peptide synthesis, crosslinking proteins to nucleic acids as well as preparation of immunoconjugates.
Water soluble condensing reagent. EDAC is generally utilized as a carboxyl activating agent for amide bonding with primary amines. In addition, it will react with phosphate groups. EDAC has been used in peptide synthesis; crosslinking proteins to nucleic acids; and preparation of immunoconjugates as examples. Typically, EDAC is utilized in the pH range 4.0-6.0 without buffers. In particular, amine and carboxylate buffers should be avoided.

Features and Benefits

Versatile and adaptable for a wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
03449E6383E7750
description

purum, ≥98.0% (AT)

description

≥99.0% (AT)

description

crystalline

description

commercial grade, powder

grade

purum

grade

-

grade

-

grade

-

assay

≥98.0% (AT)

assay

≥99.0% (AT)

assay

-

assay

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

300

application(s)

microbiology

application(s)

-

application(s)

advanced drug delivery
general analytical

application(s)

-

mp

110-115 °C

mp

112-116 °C, 110-115 °C (lit.)

mp

110-115 °C (lit.)

mp

110-115 °C (lit.)

form

powder

form

powder

form

crystalline

form

powder


Still not finding the right product?


signalword

Danger

target_organs

Stomach,large intestine,lymph node

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Hi! What is an acceptable shipping temperature, given that the storing temperature is -20C? Asking, because it came in a basic cardboard package at room temperature. And how long it can stay stable at room temp?

    1 answer
    1. This product is shipped at ambient temperature. Shipping routes are configured to minimize transit time. Freezer storage products that ship at ambient temperature are considered stable up to 2 weeks for transit time.

      Helpful?

Reviews

No rating value

Active Filters