Skip to Content
Merck
All Photos(2)

Documents

03323

Supelco

Benzo[k]fluoranthene

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12
CAS Number:
Molecular Weight:
252.31
Beilstein:
1873745
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

215-217 °C (lit.)

solubility

95% ethanol: <1 mg/mL at 20 °C
DMSO: <1 mg/mL at 20 °C
H2O: <1 mg/mL at 20 °C
acetone: 1-10 mg/mL at 20 °C
methanol: <1 mg/mL at 20 °C
toluene: 5-10 mg/mL at 20 °C

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

c1ccc2cc-3c(cc2c1)-c4cccc5cccc-3c45

InChI

1S/C20H12/c1-2-6-15-12-19-17-10-4-8-13-7-3-9-16(20(13)17)18(19)11-14(15)5-1/h1-12H

InChI key

HAXBIWFMXWRORI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Ion Mobility: TWCCSN2 value of 150.4 Å2 [M+H]+

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N 03323 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K L Willett et al.
Toxicology and applied pharmacology, 177(3), 264-271 (2001-12-26)
Certainpolynuclear aromatic hydrocarbons (PAHs) such as benzo[a]pyrene (BaP) induce CYP1A-dependent enzyme activities. Because PAHs are ubiquitous environmental contaminants, and some are aryl hydrocarbon agonists, CYP1A has been used as a biomarker for PAH exposure. However, PAHs exist in the environment
Hiroyuki Masaki et al.
The Analyst, 130(9), 1253-1257 (2005-08-13)
We developed a device and some systems for detecting benzo[a]pyrene (B[a]P) and benzo[k]fluoranthene (B[k]F). The device uses a UV light-emitting diode that emits light with a wavelength of 370 nm and a violet laser diode that emits light with a
Petra Booij et al.
Environmental toxicology and chemistry, 30(4), 898-904 (2010-12-31)
In bioassays, exposure concentrations of test compounds are usually expressed as nominal concentrations. As a result of various processes, such as adsorption, degradation, or uptake, the actual freely dissolved concentration of the test compound may differ from the nominal concentration.
Annekatrin Dreyer et al.
Environmental science & technology, 39(11), 3918-3924 (2005-06-30)
To date, studies about historic PAH (polycyclic aromatic hydrocarbons) deposition at a regional scale have rarely been published. To address this research gap, we sampled 17 ombrotrophic peatlands across eastern Canada. The peat cores from hollows were dated with 210Pb
David C Spink et al.
Toxicology and applied pharmacology, 226(3), 213-224 (2007-10-09)
The interactions of polycyclic aromatic hydrocarbons (PAH) and cytochromes P450 (CYP) are complex; PAHs are enzyme inducers, substrates, and inhibitors. In T-47D breast cancer cells, exposure to 0.1 to 1 microM benzo(k)fluoranthene (BKF) induced CYP1A1/1B1-catalyzed 17beta-estradiol (E(2)) metabolism, whereas BKF

Articles

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service