Skip to Content
Merck
All Photos(1)

Key Documents

02733

Supelco

Chromium Standard for AAS

TraceCERT®, 1 g/L Cr in nitric acid (nominal concentration)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

concentration

1 g/L Cr in nitric acid (nominal concentration)

technique(s)

AAS: suitable

application(s)

agriculture
cleaning products
cosmetics
food and beverages
personal care

format

single component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Chromium standard may have been used as reference standard in graphite furnace atomic absorption spectroscopy (GFAAS) analysis done for the determination of chromium in body fluids.

Analysis Note

1 g/L Cr in 2% nitric acid, prepared with high purity (NH4)2Cr2O7, HNO3 and water

Other Notes

For a complete product listing of our TraceCERT range of CRMs for ICP and AAS, technical information and example certificates please visit our inorganic TraceCERT website

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Met. Corr. 1 - Muta. 1B - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of chromium in whole blood and urine by graphite furnace AAS.
Huang, Y. L., et al.
Atomic Spectroscopy, 21, 10-16 (2000)
Determination of chromium in whole blood and urine by graphite furnace AAS
Huang, Y. L., et al.
Atomic Spectroscopy, 10-16 (2000)

Articles

Recent advances in analytical tech for heavy metal detection ensure food safety, with products and services supporting trust in consumption.

Recent advances in analytical tech for heavy metal detection ensure food safety, with products and services supporting trust in consumption.

Recent advances in analytical tech for heavy metal detection ensure food safety, with products and services supporting trust in consumption.

Recent advances in analytical tech for heavy metal detection ensure food safety, with products and services supporting trust in consumption.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service