Skip to Content
Merck
All Photos(1)

Key Documents

02580585

Ginkgolic acid C15:1

primary reference standard

Synonym(s):

22:1-ω7-Anacardic acid, 6-[(8Z)-Pentadecenyl]-salicylic acid, Anacardic acid monoene, Ginkgolic acid I, Romanicardic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H34O3
CAS Number:
Molecular Weight:
346.50
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

manufacturer/tradename

HWI

application(s)

food and beverages

SMILES string

CCCCCC\C=C/CCCCCCCc1cccc(O)c1C(O)=O

InChI

1S/C22H34O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-16-19-17-15-18-20(23)21(19)22(24)25/h7-8,15,17-18,23H,2-6,9-14,16H2,1H3,(H,24,25)/b8-7-

InChI key

YXHVCZZLWZYHSA-FPLPWBNLSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This compound is commonly found in plants of the genus: ginkgo

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

509.9 °F - (calculated)

Flash Point(C)

265.5 °C - (calculated)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoming Yang et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 27(1), 40-42 (2004-06-08)
To study the influence of ginkgolic acids on human tumor cells and normal cells. Ginkgolic acids (total concentration 90%) was prepared from ginkgo sarcotestas. The inhibitive effect of ginkgolic acids on human tumor cells and normal cells lines was examined
P Loungratana et al.
The American journal of Chinese medicine, 32(1), 33-48 (2004-05-25)
A competitive enzyme-linked immunosorbent assay (ELISA) for ginkgolic acids (GAs) was developed using monoclonal antibody (MAb) 9F raised against 6-(13-formylheptyl) salicylic acid covalently coupled to bovine serum albumin (BSA). ELISA, at an effective measuring range of 300 ng/ml-1 microgram/ml of
Jianming Ju et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 34(7), 817-819 (2009-07-25)
To study dynamic change of total ginkgolic acids in Ginkgo biloba leaves of the different aged trees and different collecting seasons. The content of total ginkgolic acids in G. biloba leaves was determined by HPLC. A Alltima C18 (4.6 mm
Liu-qing Yang et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 29(2), 111-115 (2005-02-22)
The latest progress in research on constituents and pharmacological activities of sarcotestas of Ginkgo biloba has been studied. The main constituents in sarcotestas of G. biloba include flavones, ginkgolides, alkylphenols, polysaccharides and amino acids, etc. They show the following activities
Xiang-yang Wu et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 38(11), 846-849 (2004-03-03)
To establish a high performance liquid chromatographic method for determination of ginkgolic acids in Ginkgo biloba extract and its preparations. Ginkgo biloba extract and its preparations were extracted with petroleum ether in Soxhlet apparatus, and then concentrated under vacuum. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service