All Photos(1)
About This Item
CAS Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
SB.52
Recommended Products
grade
for column chromatography
form
beads
technique(s)
LPLC: suitable
particle size
50-160 μm
capacity
1.0 meq/mL
bulk density
0.25 g/mL
suitability
suitable for column chromatography
separation technique
anion exchange
InChI
1S/C18H35N3O3/c19-13-7-3-5-11-17(23)21-15-9-4-6-12-18(24)20-14-8-1-2-10-16-22/h16H,1-15,19H2,(H,20,24)(H,21,23)
InChI key
HKJOYARIOKGODS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service