Skip to Content
Merck
All Photos(1)

Key Documents

01120590

Isovitexin

primary reference standard

Synonym(s):

6-C-Glucosylapigenin, Homovitexin, Saponaretin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O10
CAS Number:
Molecular Weight:
432.38
Beilstein:
66651
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

application(s)

food and beverages

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2c(O)cc3OC(=CC(=O)c3c2O)c4ccc(O)cc4

InChI

1S/C21H20O10/c22-7-14-17(26)19(28)20(29)21(31-14)16-11(25)6-13-15(18(16)27)10(24)5-12(30-13)8-1-3-9(23)4-2-8/h1-6,14,17,19-23,25-29H,7H2/t14-,17-,19+,20-,21+/m1/s1

InChI key

MYXNWGACZJSMBT-VJXVFPJBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Biochem/physiol Actions

Phytochemical found in medicinal herbs. Antioxidant.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jong Min Kim et al.
Chemistry & biodiversity, 5(2), 352-356 (2008-02-23)
Ellagic acid (1), 3,3'-di-O-methylellagic acid (2), 3,3',4-tri-O-methylellagic acid (3), isovitexin (4), kaempferol 3-O-beta-D-glucuronide methyl ester (5), quercetin 3-O-alpha-L-arabinopyranosyl-(1-->6)-beta-D-galactopyranoside (6), ursolic acid, pomolic acid, tormentic acid, euscaphic acid, euscaphic acid 28-O-beta-D-glucopyranoside, and maslinic acid were isolated from the AcOEt- and BuOH-soluble
María Inés Ragone et al.
Journal of ethnopharmacology, 113(2), 258-266 (2007-07-21)
The spasmolytic effects of an acqueous extract of cedrón (AEC) were studied on rat isolated duodenums. This plant (Aloysia citriodora Palau, Verbenaceae) is widely used for gastrointestinal disorders and as eupeptic in South America. AEC non-competitively inhibited the dose-response curve
Yujie Fu et al.
Journal of chromatography. A, 1139(2), 206-213 (2006-12-05)
Vitexin and isovitexin are a pair of isomeric compounds known as the major constituents in pigeonpea leaves and possess various pharmacological activities. In the present study, the preparative separation of vitexin and isovitexin with macroporous resins (Nankai Hecheng S &
C Y Choo et al.
Journal of ethnopharmacology, 142(3), 776-781 (2012-06-12)
The leaves of Ficus deltoidea are used as a traditional medicine by diabetes patients in Malaysia. The objective of the study is to identify and evaluate bioactive constituents with in vivo α-glucosidase inhibition. The partitioned extracts, subfractions and pure bioactive
Yujie Fu et al.
Journal of separation science, 31(2), 268-275 (2008-01-16)
A method based on ultrasonic extraction (USE) followed by LC-MS is presented for the determination of vitexin and isovitexin in pigeonpea extracts in this study. The influential parameters of the USE procedure were optimized, and the optimal conditions were as

Protocols

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service