Skip to Content
Merck

TXHSRMKB

Roche

KAPA Taq EXtra HotStart ReadyMix

suitable for PCR

Synonym(s):

KAPA, PCR, hotstart PCR, taq DNA polymerase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.55
UNSPSC Code:
41106300
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


feature

dNTPs included, hotstart

packaging

pkg of 1.25 mL (KK3604), pkg of 6.25 mL (KK3605)

manufacturer/tradename

Roche

technique(s)

PCR: suitable

input

purified DNA

storage temp.

−20°C

General description

KAPA Taq EXtra HotStart® ReadyMix with dye (2X) is a ready-to-use cocktail containing all components for short- to long-range PCR, except primers and template. The 2X ReadyMix contains KAPA Taq EXtra HotStart DNA Polymerase (0.675 U per 25 μL reaction), KAPA Taq EXtra Buffer (1X), dNTPs (0.3 mM of each dNTP at 1X), MgCl2 (2 mM at 1X) and stabilizers. The 2X ReadyMix also contains two inert tracking dyes to enable direct loading of PCR products onto agarose gels for analysis by electrophoresis, without the need to add a DNA loading solution.

Application

KAPA Taq EXtra HotStart® ReadyMix has been used for:
  • PCR of amplicons to be analyzed by agarose gel electrophoresis
  • PCR of short and medium length targets (<5 kb)
  • Long-range PCR (5 – 15 kb)
  • PCR with limiting amounts of template DNA

Features and Benefits

  • KAPA Taq EXtra HotStart® is a blend of KAPA Taq HotStart and a proofreading DNA polymerase
  • Suitable for short-, medium- and long-range PCR, and PCR with low amounts of template DNA
  • Fidelity is 2–4 times better than Taq
  • For short amplicons, replace Taq DNA polymerase for improvements in yield and/or fidelity.
  • Amplify mid- to long-range targets with high yield and sensitivity.
  • 2X ReadyMix with dye contains two inert tracking dyes to allow loading of PCR products directly onto agarose gels for analysis.

Preparation Note

Always ensure that the product has been fully thawed and mixed before use. Reagents may be stored at 4°C for short-term use (up to 1 month). Return to -20°C for long-term storage.

Analysis Note

Each batch of KAPA Taq EXtra HotStart® DNA Polymerase is confirmed to contain <2% contaminating protein (Agilent Protein 230 Assay). The 2X KAPA Taq EXtra HotStart ReadyMix is subjected to stringent quality control tests, is free of contaminating exo- and endonuclease activity, and meets strict requirements with respect to DNA contamination levels.

Other Notes

For Research Use Only. Not for use in diagnostic procedures.

Legal Information

HOTSTART is a registered trademark of Molecular BioProducts, Inc.
ReadyMix is a trademark of Sigma-Aldrich Co. LLC


Kit Components Only

Product No.
Description

  • KAPA Taq EXtra HotStart® DNA Polymerase 0.675U per 25μL reaction

  • KAPA Taq EXtra Buffer (1X)

  • 1X dNTPs 0.3 mM of each dNTP

  • 1X MgCl2 2 mM

  • Stabilizers

  • Two inert tracking dyes

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
KK360404061838087003
KK360504061837905384