Skip to Content
Merck

Skip To

TRIPURE-RO

Roche

TriPure Isolation Reagent

solution (ready-to-use), pkg of 50 mL (11667157001), pkg of 200 mL (11667165001)

Synonym(s):

DNA purification, Protein isolation, RNA isolation, nucleic acid isolation, DNA/RNA/protein extraction reagent, cell pellet RNA extraction, cell pellet RNA isolation, cell pellet RNA purification, single step RNA extraction reagent, single step RNA isolation reagent, single step RNA purification reagent, total RNA extraction solution, total RNA isolation solution, total RNA purification solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41105500
NACRES:
NA.54
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution (ready-to-use)

packaging

pkg of 200 mL (11667165001), pkg of 50 mL (11667157001)

manufacturer/tradename

Roche

storage temp.

2-8°C

General description

Monophasic solution of phenol and guanidine thiocyanate for RNA, DNA, and protein isolation.

Application

Ready-to-use reagent for the simultaneous isolation of total RNA, DNA, and proteins from cells or tissues of human, plant, yeast, bacterial, or viral origin. RNA is undegraded and free of protein or DNA contamination according to the current Quality Control.After purification, many applications are possible:
  • The DNA-free total RNA is suitable for northern blotting, in vitro translation, RNase protection assays, cDNA synthesis, or RT-PCR.[1][2][3]
  • The RNA-free DNA is suitable for PCR, restriction digestion, Southern blotting, or cloning.
  • The denatured protein is suitable for SDS-PAGE and western blotting.

Features and Benefits

The TriPure Isolation Reagent allows the simultaneous isolation of RNA, DNA, and protein from the same sample in a single-step liquid-phase separation. RNA, DNA, and protein are separated into different organic and aqueous phases, from which each can be purified by a series of alcohol precipitations.
  • Work with an easy-to-use kit. The red dye in the reagent simplifies identification of different phases.
  • Process a wide variety of starting samples. Easily adapt the kit to your specific needs.
  • Simplify isolation protocols. Use a single reagent that isolates DNA-free RNA, RNA-free DNA, and proteins.
  • Increase yields of intact RNA. TriPure Isolation Reagent provides an immediate chaotropic denaturing environment that eliminates endogenous RNase activity.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

TriPure is a trademark of Roche

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T942493289T3809
description

solution (ready-to-use), pkg of 50 mL (11667157001), pkg of 200 mL (11667165001)

description

For processing tissues, cells cultured in monolayer or cell pellets

description

-

description

BD, For processing whole blood, plasma, or serum.

manufacturer/tradename

Roche

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

-

packaging

pkg of 200 mL (11667165001)

packaging

-

packaging

-

packaging

-

form

solution (ready-to-use)

form

-

form

liquid

form

-


signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

supp_hazards

Storage Class

8A - Combustible, corrosive hazardous materials

wgk

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library