ROAMYGLL
Roche
Amyloglucosidase
from Aspergillus niger
Synonym(s):
Aspergillus niger, disaccharidase-type a-glucosidase
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
Aspergillus niger
Quality Level
form
lyophilized
specific activity
6 U/mg (approximately 6 U/mg lyophilizate at +25°C with glycogen as the substrate)
~6 units/mg protein (At 25 °C with glycogen as the substrate.)
mol wt
97 kDa
packaging
pkg of 3,500 U (11202367001)
pkg of 500 U (11202332001)
manufacturer/tradename
Roche
optimum pH
4.6-4.8
storage temp.
2-8°C
General description
Amyloglucosidase, a 1,4-α-D-glucan glucohydrolase, is a disaccharidase–type α-glucosidase. It is synthesized by several Aspergillus genus species. This exo-enzyme is one of the major industrial enzymes. The stability of amyloglucosidase can be increased by immobilization.
Specificity
Cleaves terminal glucoses that are α1,4- or α1,6-linked to an oligo- or polysaccharide of multiple glucose units. The product is D-glucose.
Application
Amyloglucosidase from Aspergillus niger is used for the determination of starch.
Biochem/physiol Actions
Amyloglucosidase from Aspergillus niger is capable of hydrolyzing the α-D-(1-4) and α-(1,6) glucosidic bonds of starch and malto-oligosaccharides. Amyloglucosidase is an extracellular enzyme that converts starch to dextrins and glucose. The enzyme is used in the starch-processing industry in the commercial production of D-glucose from corn syrup.
Preparation Note
Working concentration: Working concentration of conjugate depends on application and substrate.
The following concentrations should be taken as a guideline:
Working solution: Tris-buffered saline containing 0.1% Tween 20.
Storage conditions (working solution): 2 to 8 °C
The following concentrations should be taken as a guideline:
- Western blot: 1 to 10 μg/ml
Working solution: Tris-buffered saline containing 0.1% Tween 20.
Storage conditions (working solution): 2 to 8 °C
Other Notes
For life science research only. Not for use in diagnostic procedures.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
High fructose corn syrup: production, uses and public health concerns.
Parker K, et al.
Biotechnology and Molecular Biology Reviews, 5, 71-78 (2010)
High-potency amyloglucosidase-producing mold of the Aspergillus niger group.
Smiley K L, et al.
Applied Microbiology, 12(5), 455-455 (1964)
Simultaneous amyloglucosidase and exo-polygalacturonase production by Aspergillus niger using solid-state fermentation.
Costa, J et al.
Brazilian Archives of Biology and Technology, 50, 759-766 (2007)
Recent Advances in Basic and Applied Aspects of Industrial Catalysis (1998)
Romy Rehschuh et al.
Plant physiology, 184(2), 852-864 (2020-08-21)
Climate change increases the occurrence of prolonged drought periods with large implications for forest functioning. Scots pine (Pinus sylvestris) is one of the most abundant conifers worldwide, and evidence is rising that its resilience to severe drought is limited. However
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service