Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
biological source
Bacillus polymyxa (dispase), Bacillus sp. (thermolysin - Bacillus thermoproteolyticus), Clostridium histolyticum (collagenase)
description
high Thermolysin concentration
form
lyophilized
packaging
pkg of 10 mg (05401135001 [2 x 5 mg]), pkg of 100 mg (05401151001 [2 x 50 mg])
manufacturer/tradename
Roche
parameter
35-37 °C optimum reaction temp.
technique(s)
tissue processing: suitable
optimum pH
7.4
storage temp.
−20°C
General description
Application
Features and Benefits
- Maximize viability and yield of isolated cells
- Count on higher specific activity of the enzyme blend
- Obtain higher experimental reproducibility
- Increase safety
Preparation Note
Working concentration: Liberase Research Grade Enzyme Working Concentration:
Liberase enzymes have significantly higher specific activities than traditional collagenases. This means that the working concentration of Liberase Research Grade Purified Enzymes, expressed in mg/ml, will be lower than that of traditional collagenase.
When the application is on the Roche list of applications at www.collagenase.com, use the Liberase Research Grade concentration recommended for that application.
When the application is not included on this list, first use Liberase TM Research Grade at a concentration of 0.08–0.28 Wünsch units/ml.
The goal is to determine the best starting concentration of Liberase Research Grade Enzyme Blends. This is a starting point, and the final concentration may vary due to differences in procedure and lot-to-lot differences in traditional collagenase.
Collagenase Working Concentration:
Multiply your previous collagenase working concentration (mg/ml) by its specific activity (Wünsch units/mg, [as determined above]), to obtain Wünsch units/ml. To determine how much Liberase Research Grade Enzyme Blend to use, first multiply your collagenase working concentration (in Wünsch units/ml) times the total volume of your working enzyme solution to obtain the total collagenase activity needed (Wünsch units). Divide the total collagenase activity required by the Liberase Research Grade stock concentration ( “Reconstitution and Storage”). This indicates how many milliliters of Liberase Research Grade Enzyme Blend stock solution to use in your working enzyme solution.
Storage conditions (working solution): Store unused stock solution in single-use aliquots at -15 to -25 °C. For further information on product stability please visit the Roche Liberase Enzyme website at www.collagenase.com.
Note: Avoid repeated freezing and thawing!
Reconstitute the entire vial. Do not weigh individual aliquots of the lyophilizate. The introduction of moisture into the vial results in a decline in enzymatic activity.
Place vial on ice to rehydrate the lyophilized enzyme.
Gently agitate the vial at 2 to 8 °C until enzyme is completely dissolved (max. 30 min).
Depending on the type of tissue-dissociation buffer used to dissolve Liberase Research Grade Purified Enzyme Blends, slight precipitations may be observed which readily dissolve in the diluted working solution and have no influence on enzyme activity.
Remove an aliquot of the stock solution to prepare the working solution.
Reconstitution volume
2 ml (1 vial with 5 mg–10 mg pack size), 10 ml (1 vial with 50 mg–100 mg pack size)
Collagenase Wünsch (units/ml)
13 (1 vial with 5 mg–10 mg pack size), 26 (1 vial with 50 mg–100 mg pack size)
Total Collagenase concentration [mg/ml]
2.5 (1 vial with 5 mg–10 mg pack size), 5.0 (1 vial with 50 mg–100 mg pack size)
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) tissue processing: suitable | technique(s) single cell analysis: suitable, tissue processing: suitable | technique(s) activity assay: suitable | technique(s) tissue processing: suitable |
| biological source Bacillus polymyxa (dispase), Clostridium histolyticum (collagenase), Bacillus sp. (thermolysin - Bacillus thermoproteolyticus) | biological source - | biological source enzyme from Bacillus polymyxa (Protease), enzyme from Clostridium histolyticum (collagenase) | biological source Bacillus polymyxa (dispase), Bacillus sp. (thermolysin - Bacillus thermoproteolyticus), Clostridium histolyticum (collagenase) |
| form lyophilized | form lyophilized | form lyophilized | form lyophilized |
| optimum pH 7.4 | optimum pH 7.4 | optimum pH 7.4 | optimum pH 7.4 |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. - |
| manufacturer/tradename Roche | manufacturer/tradename Roche | manufacturer/tradename Roche | manufacturer/tradename Roche |
signalword
Danger
Hazard Classifications
Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
does not flash
flash_point_c
does not flash
hcodes
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.





