Skip to Content
Merck

Skip To

COHISC-RO

Roche

cOmplete His-Tag Purification Column

Prepacked, ready-to-use chromatography column

Synonym(s):

suitable for protein purification, matrix Sepharose-CL 6B

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.21
UNSPSC Code:
41116133
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 5 x 1 mL (columns (06781543001)), pkg of 5 mL (column (06781535001))

manufacturer/tradename

Roche

parameter

1,420 cm/hr max. flow rate

technique(s)

protein purification: suitable

matrix

Sepharose-CL 6B

capacity

≥40 mg/mL, resin binding capacity

compatibility

  • for long term storage: 20% ethanol, pH 4.0 to pH 9.0
  • during chromatography: The resin is compatible with 10mM EDTA, 10mM DTT during the purification (1 hour incubation), 6M guanidinium-HCl, 8M urea, pH 2.0 to pH 14.0.
  • during cleaning: 4% SDS for complete His-Tag Purification Resin filled in columns, pre-charged with Ni2+ stored in 20% ethanol.

storage temp.

2-8°C

General description

The most common technique in affinity purification of proteins involves engineering a sequence of 6 to 14 histidines into the N- or C-terminal (or even on an exposed loop) of the protein. Such polyhistidine stretches bind strongly to divalent metal ions such as nickel and cobalt. This effect can be used to separate proteins. Metal ions can be immobilized on a matrix using a chelator, which still allows the ion to interfere with the polyhistidine tag of the protein. When these his-tagged proteins are passed through a column containing immobilized metal ions, the proteins bind via the tag to the column. Nearly all untagged proteins pass through the column. The protein is released from the column by elution with either imidazole, which competes with the polyhistidine tag for binding to the column, or by a decrease in pH, which decreases the affinity of the tag for the resin. While this procedure is generally used for the purification of recombinant proteins with an engineered affinity tag, it can also be used for natural proteins with an inherent affinity for divalent cations.

Features and Benefits

cOmplete His-Tag Purification Columns are available in 2 sizes and prepacked with cOmplete His-Tag Purification Resin. The cOmplete His-Tag Purification Resin is an innovative high-capacity IMAC matrix (Immobilized Metal Affinity Chromatography) for convenient single-step purifications of His-tagged proteins from total lysates. Roche′s propriety nickel-chelate chemistry ensures extraordinary compatibility with commonly used reducing agents such as DTT, chelating metalloprotease inhibitors such as EDTA, and a wide range of buffer substances and salt conditions. The wide choice of compatible ingredients allows optimization of buffers for maximum protein stability and solubility.

cOmplete His-Tag Purification Columns are fully compatible with standard purification systems such as ÄKTA Systems (Cytiva).

  • Use the buffer conditions best suited to your protein
Keep your protein comfortable and let it, not your purification resin, determine whether you use DTT, EDTA, or other buffer substances.
  • Repeatedly obtain highly pure protein
Single step purification without resin recharging.
  • Protect your protein from toxic nickel
Reduce protein oxidation and aggregation caused by resins that leach nickel.
  • Work in a safe and eco-friendly environment
Avoid handling of toxic nickel and completely eliminate disposal costs.

Packaging

  • 06781535001 (5 mL column; 1 column containing 5 ml of cOmplete His-Tag Purification Resin)
  • 06781543001 (5 x 1 mL columns; 5 columns containing 1 ml of cOmplete His-Tag Purification Resin)

Other Notes

Recommended volumetric flow rate: 5 ml column (06 781 535 001): 2.5 to 10 ml/minute
1 ml column (06 781 543 001): 0.5 to 2.0 ml/minute
The volumetric flow rate is a function of the column′s cross section.

Recommended imidazole concentration for load/wash: Nonspecific binding of proteins without a His-tag is low.
Use up to 5 mM imidazole in load and/or wash buffers.
If establishing a new assay for purification of His-tag proteins with cOmplete His-Tag Purification Columns do not use imidazole. If, e.g., the purity of the His-tag protein needs to be improved following this first step, use imidazole in a final concentration of up to 5mM in a second step.

Recommended imidazole concentration for elution:
Up to 500mM
Please note:
In contrast to other available resins, bound His-tagged protein typically elutes from cOmplete His-Tag Purification Columns with a lower imidazole concentration, e.g., 25 to 45mM.

Binding capacity
The binding capacity of the resin to various types of proteins may vary according to the protein characteristics such as the size of the protein.
cOmplete His-Tag Purification Columns bind with a high specificity to the polyhistidine-tagged protein. As a consequence, the binding kinetics may appear to be different when compared to conventional metal chelate matrices. Full capacity of cOmplete His-Tag Purification Columns can be achieved by allowing more time for the protein to bind to the resin by lowering the flow rate during the chromatography purification procedure.
For life science research only. Not for use in diagnostic procedures.

Legal Information

cOmplete is a trademark of Roche

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
COHISR-ROSAB1306082HPPCRPKRO
matrix

Sepharose-CL 6B

matrix

Sepharose-CL 6B

matrix

-

matrix

-

parameter

1,420 cm/hr max. flow rate

parameter

-

parameter

-

parameter

-

capacity

≥40 mg/mL, resin binding capacity

capacity

-

capacity

-

capacity

-

technique(s)

protein purification: suitable

technique(s)

protein purification: suitable

technique(s)

western blot: 1:1000

technique(s)

DNA purification: suitable

manufacturer/tradename

Roche

manufacturer/tradename

Roche

manufacturer/tradename

-

manufacturer/tradename

Roche

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

-


signalword

Warning

Storage Class

3 - Flammable liquids

flash_point_f

93.2 °F

flash_point_c

34 °C

pictograms

FlameExclamation mark

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library